Skip to Content
Merck
All Photos(3)

Key Documents

244678

Sigma-Aldrich

Tin(IV) chloride pentahydrate

98%

Synonym(s):

Tin tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl4 · 5H2O
CAS Number:
Molecular Weight:
350.60
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
98%
form:
powder or chunks

Quality Level

Assay

98%

form

powder or chunks

reaction suitability

reagent type: catalyst
core: tin

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Sn](Cl)(Cl)Cl

InChI

1S/4ClH.5H2O.Sn/h4*1H;5*1H2;/q;;;;;;;;;+4/p-4

InChI key

KHMOASUYFVRATF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Applied in formation of a xerogel via a cyanogel the gel has potential use in solid-state gas sensors.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Idio Alves de Sousa Filho et al.
Environmental science and pollution research international, 26(5), 4224-4233 (2018-02-22)
Etoposide is an antineoplastic agent used for treating lung cancer, testicular cancer, breast cancer, pediatric cancers, and lymphomas. It is a pollutant due to its mutagenic and carcinogenic potential. Disposal of waste from this drug is still insufficiently safe, and
Xinke Liu et al.
Nanoscale research letters, 15(1), 4-4 (2020-01-05)
Polychlorinated biphenyls (PCBs) are persistent organic pollutants that are widely distributed in the environment. It is noteworthy that the PCBs are endocrine-disrupting substances, and their toxicity induces cancer and damage to the mammalian reproductive system, immune system, stomach, skin, liver
Feiyang Qiao et al.
Scientific reports, 10(1), 5024-5024 (2020-03-21)
We have successfully fabricated transparent conductive mesoporous indium tin oxide (TCM-ITO) films by a screen-printing method. The TCM-ITO films possess approximately 22 nm mesopores and obtain electrical conductivity up to 14.96 S/cm by adjusting the mass ratio of cubic-shaped ITO nanoparticles to
Ramesh Chandra et al.
Environmental science and pollution research international, 26(23), 23346-23358 (2019-06-15)
With increasing environment pollution and bacterial transmitted viral diseases globally, the development of new, effective, and low-cost materials/strategies is the current major challenge. To combat with this alarming problem, three new multi-functional and thermally stable SnO2NPs@ZIF-8 composites (NC1, NC2, and
Adsorption of cadmium ion using a new composite cation-exchanger polyaniline Sn (IV) silicate: kinetics, thermodynamic and isotherm studies.
Naushad M, et al.
International Journal of Environmental Science and Technology, 10(3), 567-578 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service