Skip to Content
Merck
All Photos(1)

Key Documents

20901

Sigma-Aldrich

Cadmium acetate dihydrate

purum p.a., ≥98.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(CH3COO)2 · 2H2O
CAS Number:
Molecular Weight:
266.53
Beilstein:
3730567
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Assay:
≥98.0% (KT)
form:
fine crystals
powder

grade

purum p.a.

Quality Level

Assay

≥98.0% (KT)

form

fine crystals
powder

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Na: ≤100 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[H]O[H].[H]O[H].CC(=O)O[Cd]OC(C)=O

InChI

1S/2C2H4O2.Cd.2H2O/c2*1-2(3)4;;;/h2*1H3,(H,3,4);;2*1H2/q;;+2;;/p-2

InChI key

AUIZLSZEDUYGDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - STOT RE 1

Target Organs

Bone,Kidney

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

113Cd and 13C chemical shielding tensors in a single crystal of cadmium (II) acetate dihydrate.
Ganapathy S, et al.
J. Chem. Phys. , 81(2), 661-668 (1984)
Mohammad Hossein Habibi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 133, 13-18 (2014-06-14)
ZnO@CdS core-shell nano-structure has been synthesized using zinc acetate dihydrate, and cadmium acetate dihydrate as simple precursors in a water-ethanol matrix without using any surfactant, ligand or chelating agents. The effect of different concentrations of cadmium acetate and sodium sulfide
Alexandra de Souza Fonseca et al.
Acta crystallographica. Section E, Structure reports online, 68(Pt 5), m635-m636 (2012-05-17)
The reaction of cadmium acetate dihydrate with 2-acetyl-pyridine (4-phenyl-thio-semicarbazone) yielded the title compound, [Cd(C(14)H(13)N(4)S)(2)]. The Cd(II) atom is six-coordin-ated in a distorted octa-hedral environment by two deproton-ated thio-semicarbazone ligands acting in a tridentate chelating mode through two N and one
Jie Yang et al.
Journal of nanoscience and nanotechnology, 12(9), 7322-7328 (2012-10-06)
CdSe/Cd(1-x) Zn(x)S core/shell quantum dots (QDs) were fabricated in 1-octadecene via a two step synthesis. CdSe cores were first prepared using CdO, trioctylphosphine (TOP) selenium, and stearic acid. Subsquently, a Cd(1-x) Zn(x)S shell coating was carried out using zinc acetate
M Lag et al.
Cell biology and toxicology, 18(1), 29-42 (2002-05-07)
The lung is a target organ for cadmium (Cd) toxicity. Apoptosis induced by cadmium acetate (CdAc) was studied in alveolar type 2 cells and Clara cells isolated from rat lung. Relatively low concentrations of CdAc (1-10 micromol/L) induced apoptosis after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service