Skip to Content
Merck
All Photos(1)

Documents

Y0001031

Lamotrigine for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Lamotrigine, 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, GI 267119X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7Cl2N5
CAS Number:
Molecular Weight:
256.09
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lamotrigine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1nnc(c(N)n1)-c2cccc(Cl)c2Cl

InChI

1S/C9H7Cl2N5/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7/h1-3H,(H4,12,13,14,16)

InChI key

PYZRQGJRPPTADH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lamotrigine for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Anticonvulsant.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacey Shim et al.
The Journal of pharmacology and experimental therapeutics, 347(2), 487-496 (2013-08-30)
Carisbamate and lamotrigine are anticonvulsants that act on neuronal voltage-gated sodium channels. Carisbamate has shown antidepressant-like effects in animal models of depression, and lamotrigine is a mood stabilizer with a therapeutic effect in depressive episodes of patients with bipolar disorder.
Shai Sandalon et al.
Experimental eye research, 115, 47-56 (2013-07-03)
Voltage gated sodium channels (Nav), are proposed mediators of neuronal damage in ischemic and excitotoxicity disease models. We evaluated the neuroprotective effects of lamotrigine, a Nav blocker, in the acute and chronic rat ocular hypertension models. Additionally, expression of the
Chaitali Ghosh et al.
Epilepsia, 54(9), 1562-1570 (2013-07-20)
Brain drug bioavailability is regulated by the blood-brain barrier (BBB). It was recently suggested that cytochrome P450 (CYP) enzymes could act in concert with multidrug transporter proteins to regulate drug penetration and distribution into the diseased brain. The possibility that
Maxine Dibué et al.
Epilepsia, 54(9), 1542-1550 (2013-06-19)
Lamotrigine (LTG) is a popular modern antiepileptic drug (AED); however, its mechanism of action has yet to be fully understood, as it is known to modulate many members of several ion channel families. In heterologous systems, LTG inhibits Cav 2.3
Allyson K Friedman et al.
Science (New York, N.Y.), 344(6181), 313-319 (2014-04-20)
Typical therapies try to reverse pathogenic mechanisms. Here, we describe treatment effects achieved by enhancing depression-causing mechanisms in ventral tegmental area (VTA) dopamine (DA) neurons. In a social defeat stress model of depression, depressed (susceptible) mice display hyperactivity of VTA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service