Skip to Content
Merck
All Photos(4)

Key Documents

T35920

Sigma-Aldrich

p-Toluenesulfonic acid monohydrate

ReagentPlus®, ≥98%

Synonym(s):

4-Methylbenzenesulfonic acid monohydrate, 4-Toluenesulfonic acid monohydrate, PTSA monohydrate, TsOH monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3H · H2O
CAS Number:
Molecular Weight:
190.22
Beilstein:
3568023
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

5.9 (vs air)
5.9 (vs air)

Quality Level

product line

ReagentPlus®

Assay

≥98%

form

solid

mp

103-106 °C ((lit.))
103-106 °C (lit.)

functional group

tosylate

SMILES string

[H]O[H].Cc1ccc(cc1)S(O)(=O)=O

InChI

[1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2]
1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2

InChI key

[KJIFKLIQANRMOU-UHFFFAOYSA-N]
KJIFKLIQANRMOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

p-toluenesulfonic acid monohydrate may be used as a better alternative to Friedel-Crafts catalysts for the alkylation of the aromatic nucleus with activated alkyl halides, alkenes, or tosylates under mild conditions.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detailed characterization of p-toluenesulfonic acid monohydrate as a convenient, recoverable, safe, and selective catalyst for alkylation of the aromatic nucleus.
Mahindaratne MPD and Wimalasena K.
The Journal of Organic Chemistry, 63(9), 2858-2866 (1998)
Hans H Adomat et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 902, 84-95 (2012-07-24)
Androgens are key mediators of prostate development and function, a role that extends to the development of prostate diseases such as benign prostatic hyperplasia (BPH) and prostate cancer. In prostate, DHT is the major androgen and reduction and glucuronidation are
Mild C-H halogenation of anilides and the isolation of an unusual palladium(I)-palladium(II) species.
Robin B Bedford et al.
Angewandte Chemie (International ed. in English), 50(24), 5524-5527 (2011-05-14)
G Shanmugam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 177-183 (2012-05-29)
Bulk crystals of newly identified organic nonlinear optical material 4-methylanilinium p-toluenesulfonate (PMPT) were grown by slow evaporation solution growth method using ethanol as a solvent. It crystallizes in monoclinic system with a noncentrosymmetric space group P2(1). The formation of the
Pei Liu et al.
Organic & biomolecular chemistry, 10(24), 4696-4698 (2012-05-18)
A convenient one-pot Mannich-type-cyclization-oxidation tandem process has been developed for the synthesis of 1,3,5-trisubstituted pyrazoles derivatives from aldehydes, hydrazines and alkynes using p-toluenesulfonic acid monohydrate (PTSA) as a multifunctional catalyst. This method provides a flexible and rapid route to 1,3,5-trisubstituted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service