Skip to Content
Merck
All Photos(3)

Key Documents

STS0215

Sigma-Aldrich

ECO Brij® S10

greener alternative

average Mn ~711

Synonym(s):

Brij® S10, Polyethylene glycol octadecyl ether, Polyoxyethylene (10) stearyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H37(OCH2CH2)nOH, n~10
CAS Number:
MDL number:
UNSPSC Code:
12165104
NACRES:
NA.28

description

non-ionic

Quality Level

form

solid

mol wt

average Mn ~711

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

37-39 °C (lit.)

solubility

ethanol: 0.1%, clear, colorless

density

0.964 g/mL at 25 °C (lit.)

HLB

12.4

greener alternative category

InChI

1S/C20H41O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h2-20H2,1H3

InChI key

PSEGVHKUPXNGGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ECO Brij® S10 is an ethoxylated natural fatty alcohol, a nonionic bio-based surfactant that belongs to the class of polyoxyethylene ethers. This naturally derived detergent offers a number of functional benefits including detergency, wetting and dispersing making it suitable for a range of applications in biochemical and biological research.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biobased surfactant and is aligned with the 7th principle of Green Chemistry "Use of Renewable Feedstocks" and the 10th principle "Design for Degradation".

Features and Benefits

  • 100 % Renewable
  • 100 % Bio-based
  • Certified to the USDA BioPreferred Program
  • Lower carbon footprint than petrochemical-based versions
  • High-purity chemical suitable for a wide variety of research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Brij is a registered trademark of Croda International PLC
ECO BRIJ is a registered trademark of Croda Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingyan Song et al.
Journal of colloid and interface science, 328(2), 374-384 (2008-10-17)
Patternable nanoporous silica thin films with pore sizes on multiple length scales are fabricated using preformed block copolymer/homopolymer blend films as templates. Previous work by Tirumala et al. [V.R. Tirumala, R.A. Pai, S. Agarwal, J.J. Testa, G. Bhatnagar, A.H. Romang
Zhengrong Cui et al.
The Journal of pharmacy and pharmacology, 54(9), 1195-1203 (2002-10-03)
Intranasal immunization offers potential forthe elicitation of effective mucosal and systemic immune responses. In this study, a previously reported novel cationic nanoparticle engineered from a microemulsion precursor was further modified, optimized and applied intranasally to mice to explore its potential
S Rahima Benhabbour et al.
Journal of controlled release : official journal of the Controlled Release Society, 158(1), 63-71 (2011-11-01)
Lipid-based oil-filled nanoparticles (NPs) with a high concentration of surface-chelated nickel (Ni-NPs) were successfully prepared using a Brij 78-NTA-Ni conjugate synthesized with Brij 78 (Polyoxyethylene (20) stearyl ether) and nitrilotriacetic acid (NTA). The facile incorporation of the Brij 78-NTA-Ni conjugate
Christian Klammt et al.
Journal of structural biology, 158(3), 482-493 (2007-03-14)
G-protein coupled receptors (GPCRs) are key elements in signal transduction pathways of eukaryotic cells and they play central roles in many human diseases. So far, most structural and functional approaches have been limited by the immense difficulties in the production
Highly enantioselective epoxidation of 2,4-diarylenones by using dimeric cinchona phase-transfer catalysts: enhancement of enantioselectivity by surfactants.
Sang-Sup Jew et al.
Angewandte Chemie (International ed. in English), 44(9), 1383-1385 (2005-01-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service