Skip to Content
Merck
All Photos(1)

Documents

S1950000

Sulfadoxine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sulfadoxin, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide, Sulfadoxine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O4S
CAS Number:
Molecular Weight:
310.33
Beilstein:
625453
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfadoxine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ncnc(NS(=O)(=O)c2ccc(N)cc2)c1OC

InChI

1S/C12H14N4O4S/c1-19-10-11(14-7-15-12(10)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

PJSFRIWCGOHTNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfadoxine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexia Daoust et al.
NeuroImage, 96, 133-142 (2014-04-08)
The MAP6 (microtubule-associated protein 6) KO mouse is a microtubule-deficient model of schizophrenia that exhibits severe behavioral disorders that are associated with synaptic plasticity anomalies. These defects are alleviated not only by neuroleptics, which are the gold standard molecules for
Sankar Sridaran et al.
The American journal of tropical medicine and hygiene, 90(5), 840-845 (2014-03-13)
Chloroquine (CQ) is used as a first-line therapy for the treatment of Plasmodium falciparum malaria in Nicaragua. We investigated the prevalence of molecular markers associated with CQ and sulfadoxine-pyrimethamine (SP) resistance in P. falciparum isolates obtained from the North Atlantic
C H Sibley et al.
Trends in parasitology, 17(12), 582-588 (2002-01-05)
Chemotherapy remains the only practicable tool to control falciparum malaria in sub-Saharan Africa, where >90% of the world's burden of malaria mortality and morbidity occurs. Resistance is rapidly eroding the efficacy of chloroquine, and the combination pyrimethamine-sulfadoxine is the most
H M McIntosh et al.
Annals of tropical medicine and parasitology, 92(3), 265-270 (1998-08-26)
The 4-aminoquinolines, chloroquine or amodiaquine, have long been the drugs of choice for treating uncomplicated, falciparum malaria in Africa, although resistance to them is now common. Sulfadoxine-pyrimethamine (SP) is the usual alternative when 4-aminoquinoline treatment fails. A combination-treatment regimen could
Philip J Peters et al.
Drug safety, 30(6), 481-501 (2007-06-01)
Plasmodium falciparum infection during pregnancy is strongly associated with maternal anaemia and low birth weight, contributing to substantial morbidity and mortality in sub-Saharan Africa. Intermittent preventive treatment in pregnancy with sulfadoxine/pyrimethamine (IPTp-SP) has been one of the most effective approaches

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service