Skip to Content
Merck
All Photos(1)

Documents

S0040000

Saccharin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,3-Dihydroxy-1,2-benzisothiazol-3-one-1,1-dioxide, 2-Sulfobenzoic acid imide, o-Benzoic sulfimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO3S
CAS Number:
Molecular Weight:
183.18
Beilstein:
6888
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

saccharin

manufacturer/tradename

EDQM

mp

226-229 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1NS(=O)(=O)c2ccccc12

InChI

1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)

InChI key

CVHZOJJKTDOEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Saccharin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

A sweet tastant for mammals. A glycerol taste receptor binding site specific for glucose has been proposed in drosophila.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Whysner et al.
Pharmacology & therapeutics, 71(1-2), 225-252 (1996-01-01)
Sodium saccharin (NaSac) produces bladder tumors consistently in male rats only after lifetime exposure that begins at birth. NaSac is not metabolized and is negative in most genotoxicity tests. NaSac-induced cell damage and proliferation have been proposed as important factors
The toxicological effects of saccharin in short-term genotoxicity assays.
D L Arnold et al.
Mutation research, 221(2), 69-132 (1989-09-01)
D L Arnold
Fundamental and applied toxicology : official journal of the Society of Toxicology, 4(5), 674-685 (1984-10-01)
Saccharin, like most of the artificial sweeteners, was discovered quite by accident. The controversy regarding its use as a food additive is reflected by the number of chronic/carcinogenic studies conducted to assess its safety. Two chronic studies were conducted prior
R L Anderson
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 26(7), 637-644 (1988-07-01)
An hypothesis is presented of a mechanism for the sodium saccharin (NaS)-associated tumorigenesis of the urinary bladder that occurs in male rats. The ingestion of high doses of NaS is associated with increased urine volume and bladder mass. In rats
A G Renwick
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 23(4-5), 429-435 (1985-04-01)
Recent studies on saccharin in animals and man have allowed a detailed understanding of its fate in the body. Saccharin is slowly absorbed from the gut but rapidly eliminated in the urine, largely by renal tubular secretion. Saccharin does not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service