Skip to Content
Merck
All Photos(1)

Documents

PHR1757

Supelco

Nevirapine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Nevirapine, 11-Cyclopropyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2′,3′-e][1,4]diazepin-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N4O
CAS Number:
Molecular Weight:
266.30
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000520
traceable to USP 1460703

API family

nevirapine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CC1=CC=NC2=C1NC(C(C=CC=N3)=C3N2C4CC4)=O

InChI

1S/C15H14N4O/c1-9-6-8-17-14-12(9)18-15(20)11-3-2-7-16-13(11)19(14)10-4-5-10/h2-3,6-8,10H,4-5H2,1H3,(H,18,20)

InChI key

NQDJXKOVJZTUJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Nevirapine is a non-nucleoside reverse transcriptase inhibitor (NNRTI), which belongs to the class of antiretroviral drugs. It acts by inhibiting the reverse transcriptase enzyme, thus suppressing the multiplication of HIV. It is highly effective against the advanced HIV disease.

Application

Nevirapine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and fixed dose combinations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Nevirapine is an allosteric, non-nucleoside inhibitor of HIV reverse transcriptase (NNRTI). The Ki for inhibition of wild-type RT by Nevirapine is 200 nM.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8118 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

MEKC determination of antiretroviral reverse transcriptase inhibitors lamivudine, stavudine, and nevirapine in pharmaceutical formulations
Sekar R and Azhaguvel S
Chromatographia, 67(5-6), 389-398 (2008)
Development and validation of a simple and rapid capillary zone electrophoresis method for determination of nnrti nevirapine in pharmaceutical formulations
Zanolli F, et al.
Journal of the Brazilian Chemical Society, 22(10), 2005-2012 (2011)
Nevirapine
USP42-NF37
United States Pharmacopeia/National Formulary, 32(3), 3101-3101 (2018)
Simultaneous determination of lamivudine, stavudine and nevirapine in antiretroviral fixed dose combinations by high performance liquid chromatography
Kapoor N, et al.
Analytica Chimica Acta, 570(1), 41-45 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service