Skip to Content
Merck
All Photos(1)

Key Documents

PHR1140

Supelco

Ketorolac Tromethamine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ketorolac tris salt, (±)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid tris salt, Toradol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO3 · C4H11NO3
CAS Number:
Molecular Weight:
376.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000486
traceable to USP 1356665

API family

ketorolac

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

NC(CO)(CO)CO.OC(=O)C1CCn2c1ccc2C(=O)c3ccccc3

InChI

1S/C15H13NO3.C4H11NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13;5-4(1-6,2-7)3-8/h1-7,11H,8-9H2,(H,18,19);6-8H,1-3,5H2

InChI key

BWHLPLXXIDYSNW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Ketorolac Tromethamine belongs to the class of non-steroidal anti-inflammatory drugs, and its mode of action involves the inhibition of the bodily synthesis of prostagladins. It exhibits anti-inflammatory, antipyretic and analgesic effects.[1]

Application

Ketorolac Tromethamine may be used pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations[1][2] using reversed-phase high-performance liquid chromatography[1] and spectrophotometric technique.[2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7191 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A validated RP-HPLC method for simultaneous estimation of febuxostat and ketorolac tromethamine in pharmaceutical formulations
Gandla K, et al.
Journal of Drug Delivery and Therapeutics, 2(3) (2012)
Simultanious spectrophotometric estimation of ofloxacin and ketorolac tromethamine in ophthalmic dosage form
Fegade DJ, et al.
International Journal of ChemTech Research, 1(3), 189-194 (2009)
Giuseppina Magni et al.
Anesthesia and analgesia, 116(2), 443-447 (2013-01-11)
Ketorolac tromethamine (ketorolac) is a nonsedating drug with potent analgesic and moderate anti-inflammatory activity, which does not increase the sedation level. The safety of ketorolac with respect to risk of bleeding has been demonstrated in large numbers of patients undergoing
Stephen J Kim et al.
Archives of ophthalmology (Chicago, Ill. : 1960), 130(4), 456-460 (2012-04-12)
To investigate the adverse ocular effects of intravitreal ketorolac (4 mg) in patients with chronic uveitis and complications of chronic inflammation (macular edema). We conducted a prospective phase 1 clinical trial involving 10 eyes of 10 adult patients with chronic
Michel Weber et al.
Acta ophthalmologica, 91(1), e15-e21 (2012-09-14)
To determine whether indomethacin 0.1% eye drops are at least as effective as ketorolac 0.5% eye drops in treating ocular inflammation following cataract surgery. Prospective, multicenter, investigator-masked, parallel-group, randomized, active-controlled clinical trial. Cataract patients were randomized in a 1:1 ratio

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service