Skip to Content
Merck
All Photos(1)

Key Documents

NIST895

Stainless Steel

NIST® SRM® 895, (SAE 201)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 150 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

SRM chips range between sizes 0.50 and 1.18mm, corresponding to sieve openings of 35 and 16 mesh.



SRM 895_cert

SRM 895_SDS

Application

Stainless steel SRM is primarily intended to be used for chemical analysis methods.

Features and Benefits

Accompanied with certificate documenting both NIST-certified and non-certified values of elements present in stainless steel.

Other Notes

Certified for the analytes listed below.
Carbon (C), Manganese (Mn), Phosphorous (P), Sulfur (S), Silicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Molybdenum (Mo), Cobalt (Co)

Reference/Informational value is provided for the following analytes.
Aluminium (Al), Antimony (Sb), Arsenic (As), Bismuth (Bi), Boron (B), Lead (Pb), Niobium (Nb), Selenium (Se), Tantalum (Ta), Tellurium (Te), Tin (Sn), Titanium (Ti), Tungsten (W)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service