Skip to Content
Merck
All Photos(1)

Key Documents

E2499900

Etofenamate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-[[3-(Trifluoromethyl)phenyl]amino]benzoic acid 2-(2-hydroxyethoxy)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18F3NO4
CAS Number:
Molecular Weight:
369.34
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

etofenamate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C18H18F3NO4/c19-18(20,21)13-4-3-5-14(12-13)22-16-7-2-1-6-15(16)17(24)26-11-10-25-9-8-23/h1-7,12,22-23H,8-11H2

InChI key

XILVEPYQJIOVNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Etofenamate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Action spectrum for etofenamate photoallergic contact dermatitis.
Alastair Kerr et al.
Contact dermatitis, 65(2), 117-118 (2011-07-13)
G Vanderstraeten et al.
Acta Belgica. Medica physica : organe officiel de la Societe royale belge de medecine physique et de rehabilitation, 13(3), 139-141 (1990-07-01)
In this 60 patient study of sports traumatology due to football injuries, etofenamate gel proved equally effective as oral naproxen on the overall pain scores (65% none to mild pain for etofenamate versus 86% for naproxen; p greater than 0.05).
Meral Şen et al.
Digestive diseases and sciences, 54(12), 2742-2747 (2009-01-02)
Pain and diaphragmatic dysfunction are the major reasons for postoperative pulmonary complications after upper abdominal surgery. Preoperative administration of analgesics helps to reduce and prevent pain. The objective of this study was first to research the rate of pulmonary complications
Contact dermatitis due to etofenamate.
L Guerra et al.
Contact dermatitis, 26(3), 199-199 (1992-03-01)
Allergic contact dermatitis from etofenamate: report of 9 cases.
J P Hergueta et al.
Contact dermatitis, 31(1), 60-62 (1994-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service