Skip to Content
Merck
All Photos(1)

Key Documents

C2248000

Clenbuterol hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Amino-α-(t-butylaminomethyl)-3,5-dichlorobenzyl alcohol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18Cl2N2O · HCl
CAS Number:
Molecular Weight:
313.65
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clenbuterol

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CC(C)(C)NCC(O)c1cc(Cl)c(N)c(Cl)c1

InChI

1S/C12H18Cl2N2O.ClH/c1-12(2,3)16-6-10(17)7-4-8(13)11(15)9(14)5-7;/h4-5,10,16-17H,6,15H2,1-3H3;1H

InChI key

OPXKTCUYRHXSBK-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clenbuterol hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Vulić et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 77, 82-92 (2015-01-18)
The aim of this study was to evaluate in vitro toxicity of clenbuterol and its metabolite 4-amino-3,5-dichlorobenzoic acid. Cytotoxicity and pro-oxidative effect of both compounds were studied on human colon adenocarcinoma cell line SW 480. No significant cytotoxic effect of
Jiandong Hu et al.
PloS one, 10(3), e0122005-e0122005 (2015-03-24)
A surface plasmon resonance (SPR) immunoassay with an immobilization of self-assembled molecular identification membrane for the detection of residual Clenbuterol Hydrochloride (CLB) in pork liver was systematically investigated and experimentally validated for its high performance. SPR immunoassay with a regular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service