Skip to Content
Merck
All Photos(1)

Key Documents

C0250000

Calcium folinate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Folinic acid calcium salt hydrate, 5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt, 5-Formyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid calcium salt, 5-HCO-H4PteGlu, Calcium folinate, Citrovorum factor calcium salt, Leucovorin calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21CaN7O7 · xH2O
CAS Number:
Molecular Weight:
511.50 (anhydrous basis)
Beilstein:
5723924
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

folic acid

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Ca++].[H]O[H].[H]C(=O)N1C(CNc2ccc(cc2)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)CNc3nc(N)nc(O)c13

InChI

1S/C20H23N7O7.Ca/c21-20-25-16-15(18(32)26-20)27(9-28)12(8-23-16)7-22-11-3-1-10(2-4-11)17(31)24-13(19(33)34)5-6-14(29)30;/h1-4,9,12-13,22H,5-8H2,(H,24,31)(H,29,30)(H,33,34)(H4,21,23,25,26,32);/q;+2/p-2/t12?,13-;/m0./s1

InChI key

KVUAALJSMIVURS-ZEDZUCNESA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia. Calcium folinate (CF) is a folic acid derivative. It is a naturally occurring compound that has a vital role in various metabolic pathways. Calcium folinate (CF) drug is also used in the treatment of anemia, during pregnancy.[1]

Application

Calcium folinate offsets the effects of folic acid antagonists that act by binding dihydrofolate reductase and blocking the conversion of folic acid to tetrahydrofolate. In medicine, calcium folinate is used after treatment with methotrexate to reduce the toxicity of the methotrexate. Calcium folinate is also used to enhance the cytotoxic effect of 5-fluorouracil in the treatment of cancer.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC- MS characterization of trace impurities contained in calcium folinate
Francese G, et al.
Journal of Pharmaceutical and Biomedical Analysis, 39(3-4), 757-763 (2005)
Harpreet Wasan et al.
The Lancet. Oncology, 15(6), 631-639 (2014-04-08)
Advanced colorectal cancer is treated with a combination of cytotoxic drugs and targeted treatments. However, how best to minimise the time spent taking cytotoxic drugs and whether molecular selection can refine this further is unknown. The primary aim of this
Akira Ooki et al.
Japanese journal of clinical oncology, 44(4), 383-387 (2014-02-22)
We have planned a multicentre prospective study to examine the relative impact of the efficacy and adverse events of cetuximab plus first-line chemotherapy on the quality of life in Japanese patients with KRAS wild-type unresectable colorectal cancer. The Dermatology Life
High cut-off dialysis as a salvage therapy option in high-dose methotrexate chemotherapy?
Anna Bertram et al.
Annals of hematology, 93(6), 1053-1055 (2013-10-08)
Na Yeon Han et al.
Radiology, 271(2), 416-425 (2014-01-31)
To retrospectively evaluate findings of chemotherapy-induced focal hepatopathy (CIFH) on gadoxetic acid-enhanced magnetic resonance (MR) and diffusion-weighted (DW) images and to determine imaging features that are most helpful in differentiating CIFH from metastasis. This retrospective study was approved by the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service