Skip to Content
Merck
All Photos(1)

Key Documents

B1385

Supelco

Butoxamine hydrochloride

analytical standard, for drug analysis, mixture of diastereomers

Synonym(s):

α-(1-[t-Butylamino]ethyl)-2,5-dimethoxybenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H25NO3 · HCl
CAS Number:
Molecular Weight:
303.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.COc1ccc(OC)c(c1)C(O)C(C)NC(C)(C)C

InChI

1S/C15H25NO3.ClH/c1-10(16-15(2,3)4)14(17)12-9-11(18-5)7-8-13(12)19-6;/h7-10,14,16-17H,1-6H3;1H

InChI key

URPAECSKKQLCII-UHFFFAOYSA-N

General description

Butoxamine hydrochloride is a β-andrenoceptor antagonist, which shows β2-selectivity.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nozomu Abe et al.
BioMed research international, 2020, 3214186-3214186 (2020-05-29)
Adrenaline quickly inhibits the release of histamine from mast cells. Besides β2-adrenergic receptors, several in vitro studies also indicate the involvement of α-adrenergic receptors in the process of exocytosis. Since exocytosis in mast cells can be detected electrophysiologically by the
Claudio Molinari et al.
Endocrinology, 148(8), 4080-4090 (2007-04-28)
Prolactin has been associated with many effects and has been implicated in the pathogenesis of pregnancy-related hypertensive disorders, although little is known about its vascular effects. The present study was designed to determine the primary effect of prolactin on regional
Jiayin Lu et al.
International journal of molecular sciences, 22(3) (2021-02-06)
Restraint stress causes various maternal diseases during pregnancy. β2-Adrenergic receptor (β2-AR) and Forkhead transcription factor class O 1 (FOXO1) are critical factors not only in stress, but also in reproduction. However, the role of FOXO1 in restraint stress, causing changes
Cardiovascular teratogenicity of terbutaline and ritodrine in the chick embryo
Lenselink R.D, et al.
American Journal of Obstetrics and Gynecology, 171(2), 501-506 (1994)
Concise Dictionary of Pharmacological Agents: Properties and Synonyms null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service