Skip to Content
Merck
All Photos(6)

Documents

A5131

Sigma-Aldrich

L-Arginine monohydrochloride

≥98% (HPLC)

Synonym(s):

(S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
Beilstein:
3631658
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Arginine monohydrochloride, reagent grade, ≥98% (HPLC), powder

grade

reagent grade

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

mp

>235 °C

solubility

H2O: 50 mg/mL

application(s)

cell analysis

SMILES string

Cl[H].N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1

InChI key

KWTQSFXGGICVPE-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Arginine monohydrochloride has been used to study the effects of amino acids in promoting food consumption in Drosophila melanogaster. It has also been used to develop a non-invasive chronic pancreatitis model of rat.

Biochem/physiol Actions

L-Arginine is a semi-essential amino acid. This dibasic amino acid acts as a precursor for creatinine and is a natural constituent of most of the dietary proteins.
Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Severe Chronic Pancreatitis due to Recurrent Acute Injury: Non-Invasive Chronic Pancreatitis Model of Rat
Surendra Sharma
Pancreas (2016)
J. P. Felix D'Mello
Amino Acids in Human Nutrition and Health (2012)
An internal sensor detects dietary amino acids and promotes food consumption in Drosophila
Zhe Yang
Nature (2017)
Enyong Dai et al.
Nature communications, 11(1), 6339-6339 (2020-12-15)
Ferroptosis is a more recently recognized form of cell death that relies on iron-mediated oxidative damage. Here, we evaluate the impact of high-iron diets or depletion of Gpx4, an antioxidant enzyme reported as an important ferroptosis suppressor, in the pancreas
Fan Zeng et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 374(1784), 20190197-20190197 (2019-09-10)
Tunicates populate a great variety of marine underwater substrates worldwide and represent a significant concern in marine shipping and aquaculture. Adhesives are secreted from the anterior papillae of their swimming larvae, which attach and metamorphose into permanently adhering, filter-feeding adults.

Articles

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service