Skip to Content
Merck
All Photos(1)

Documents

A3278

Supelco

D-Amphetamine hemisulfate salt solution

1.0 mg/mL in methanol, analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N · 0.5 H2SO4
CAS Number:
Molecular Weight:
184.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

drug control

Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.C[C@H](N)Cc1ccccc1.C[C@H](N)Cc2ccccc2

InChI

1S/2C9H13N.H2O4S/c2*1-8(10)7-9-5-3-2-4-6-9;1-5(2,3)4/h2*2-6,8H,7,10H2,1H3;(H2,1,2,3,4)/t2*8-;/m00./s1

InChI key

PYHRZPFZZDCOPH-QXGOIDDHSA-N

Gene Information

human ... SLC18A2(6571)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Scott F Owen et al.
Nature neuroscience, 22(7), 1061-1065 (2019-06-19)
A key assumption of optogenetics is that light only affects opsin-expressing neurons. However, illumination invariably heats tissue, and many physiological processes are temperature-sensitive. Commonly used illumination protocols increased the temperature by 0.2-2 °C and suppressed spiking in multiple brain regions. In
Lucas E Cromberg et al.
Journal of neurochemistry, 149(3), 362-380 (2019-01-22)
The process of locomotion is controlled by fine-tuned dopaminergic neurons in the Substantia Nigra pars-compacta (SNpc) that projects their axons to the dorsal striatum regulating cortical innervations of medium spiny neurons. Dysfunction in dopaminergic neurotransmission within the striatum leads to
Jakob William von Trotha et al.
The European journal of neuroscience, 40(9), 3302-3315 (2014-08-26)
The brain reward circuitry plays a key role in emotional and motivational behaviors, and its dysfunction underlies neuropsychiatric disorders such as schizophrenia, depression and drug addiction. Here, we characterized the neuronal activity pattern induced by acute amphetamine administration and during
Heather N Lavezzi et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(3), 676-687 (2014-08-29)
The rostromedial tegmental nucleus (RMTg) is a strong inhibitor of dopamine neurons in the ventral tegmental area (VTA) reported to influence neurobiological and behavioral responses to reward omission, aversive and fear-eliciting stimuli, and certain drugs of abuse. Insofar as previous

Protocols

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Chromatograms

suitable for GCapplication for HPLCapplication for HPLCsuitable for GCShow More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service