Skip to Content
Merck
All Photos(1)

Key Documents

A0230000

Adenine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6-Aminopurine, Vitamin B4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
Beilstein:
5777
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

adenine

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

>360 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ncnc2[nH]cnc12

InChI

1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)

InChI key

GFFGJBXGBJISGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia. Adenine belongs to the group of purine alkaloids widely distributed in plant products and beverages. It can be used as an internal microbial marker for the determination of microbial protein synthesis.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of adenine, caffeine, theophylline and theobromine by HPLC with amperometric detection
Meyer A, et al.
Fresenius Journal of Analytical Chemistry, 356(3-4), 284-287 (1996)
Simultaneous determination of adenine and guanine in ruminant bacterial pellets by ion-pair HPLC
Moral dGP, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 826(1-2), 257-260 (2005)
Michinori Matsuo et al.
Journal of molecular and cellular cardiology, 38(6), 907-916 (2005-05-25)
ATP-sensitive potassium (K(ATP)) channels are regulated by adenine nucleotides to convert changes in cellular metabolic levels into membrane excitability. Hence, elucidation of interaction of SUR and Kir6.x with adenine nucleotides is an important issue to understand the molecular mechanisms underlying
Didier Wion et al.
Nature reviews. Microbiology, 4(3), 183-192 (2006-02-21)
N(6)-methyl-adenine is found in the genomes of bacteria, archaea, protists and fungi. Most bacterial DNA adenine methyltransferases are part of restriction-modification systems. Certain groups of Proteobacteria also harbour solitary DNA adenine methyltransferases that provide signals for DNA-protein interactions. In gamma-proteobacteria
Georg Behrens et al.
AIDS patient care and STDs, 28(4), 168-175 (2014-03-26)
The once daily, single-tablet regimen (STR) combining rilpivirine (RPV), emtricitabine (FTC), and tenofovir disoproxil fumarate (TDF) provides a simplified treatment option for antiretroviral therapy (ART)-naïve patients with baseline HIV-1 RNA (BLVL) of ≤100,000 copies/mL. The aim of this analysis is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service