Skip to Content
Merck
All Photos(2)

Key Documents

94496

Supelco

Carbamazepine

analytical standard

Synonym(s):

5H-Dibenz[b,f]azepine-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O
CAS Number:
Molecular Weight:
236.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

impurities

≤0.5% water

mp

191-192 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)N1c2ccccc2C=Cc3ccccc13

InChI

1S/C15H12N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H,(H2,16,18)

InChI key

FFGPTBGBLSHEPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Anticonvulsant; ligand for the GABAA receptor benzodiazepine modulatory site. Sodium channel inhibitor.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ursula Amstutz et al.
Epilepsia, 55(4), 496-506 (2014-03-07)
To systematically review evidence on genetic risk factors for carbamazepine (CBZ)-induced hypersensitivity reactions (HSRs) and provide practice recommendations addressing the key questions: (1) Should genetic testing for HLA-B*15:02 and HLA-A*31:01 be performed in patients with an indication for CBZ therapy
V L Yip et al.
Clinical pharmacology and therapeutics, 92(6), 757-765 (2012-11-08)
Carbamazepine (CBZ) therapy is associated with cutaneous adverse reactions in up to 10% of patients. Predisposition to these hypersensitivity reactions has been linked to the human leukocyte antigen (HLA) genotype. This systematic review determines the strength of these associations and
Janneke Jentink et al.
BMJ (Clinical research ed.), 341, c6581-c6581 (2010-12-04)
To identify specific major congenital malformations associated with use of carbamazepine in the first trimester of pregnancy. A review of all published cohort studies to identify key indications and a population based case-control study to test these indications. Review of
Sandeep Grover et al.
Pharmacogenetics and genomics, 24(2), 94-112 (2013-12-18)
A considerable heterogeneity exists in the literature on the role of different HLA alleles in carbamazepine (CBZ)-induced cutaneous adverse drug reactions (cADRs) of varying severity among diverse ethnic groups. The aim of the present study was to understand and summarize
Robert J Hatch et al.
Epilepsia, 55(4), 601-608 (2014-03-13)
A NaV β1(C121W) mouse model of human genetic epilepsy has enhanced neuronal excitability and temperature sensitivity attributed to a decreased threshold for action potential firing in the axon initial segment. To investigate the network consequences of this neuronal dysfunction and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service