Skip to Content
Merck
All Photos(1)

Key Documents

79893

Millipore

Baird-Parker Agar

NutriSelect® Plus

Synonym(s):

BPA (RPF), Baird Parker (RPF) Agar, RPF = Rabbit Plasma Fibrinogen, Baird Parker agar base (RPF)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

Agency

ISO 6888-1:2020
ISO 6888-2:2020

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 20.0 g/L
casein enzymic hydrolysate, 10.0 g/L
glycine, 12.0 g/L
lithium chloride, 5.0 g/L
meat extract, 5.0 g/L
Rabbit Plasma Fibrinogen
sodium pyruvate, 10.0 g/L
yeast extract, 1.0 g/L

packaging

pkg of 500 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.2±0.2 (25 °C)

application(s)

bioburden testing
clinical testing
environmental
food and beverages
microbiology

suitability

Staphylococcus spp.

Application

Baird Parker Agar Base is recommended for the isolation and enumeration of coagulase positive Staphylococci from food and other materials using RPF Supplement (Cat. No. 05939).

Preparation Note

Suspend 6.3 grams in 90 ml distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool to 50°C and aseptically add rehydrated content of 1 vial RPF Supplement (Cat. No. 05939). Mix well and pour into sterile petri plates.Warning : Lithium Chloride is harmful. Avoid all bodily contact and inhalation of vapours. On contact with skin wash with plenty of water immediately.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service