Skip to Content
Merck
All Photos(1)

Key Documents

76511

Supelco

Resveratrol

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,4′,5-Trihydroxy-trans-stilbene, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)\C=C\c2cc(O)cc(O)c2

InChI

1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H/b2-1+

InChI key

LUKBXSAWLPMMSZ-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Resveratrol (RV) is a polyphenol nonflavonoid compound mainly found in grapes and red wine and also in some strong pigmented fruits and vegetables that exhibits antitumor, antioxidant, antiviral, and phytoestrogenic biological activities.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Resveratrol (RV) has been used as a reference standard for the determination of trans- and cis-resveratrol in wine by capillary electrophoresis method.
Selective inhibitor of COX−1.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>392.0 °F - closed cup - (External MSDS)

Flash Point(C)

> 200.0 °C - closed cup - (External MSDS)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therapeutic potential of resveratrol: the in vivo evidence.
Baur JA, et al.
Nature Reviews. Drug Discovery, 5(6), 493-493 (2006)
Multiplicity of effects and health benefits of resveratrol.
Kursvietien? L, et al.
Medicina (Kaunas, Lithuania), 52(3), 148-155 (2016)
Capillary electrophoresis analysis of trans-and cis-resveratrol, quercetin, catechin and gallic acid in wine.
Prasongsidh B C, et al.
Food Chemistry, 62(3), 355-358 (1998)
Satya Dash et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(12), 2895-2901 (2013-09-28)
Overproduction of hepatic apolipoprotein B (apoB)-100 containing very low-density lipoprotein particles and intestinal apoB-48 containing chylomicrons contributes to hypertriglyceridemia seen in conditions such as obesity and insulin resistance. Some, but not all, preclinical and clinical studies have demonstrated that the
Alexandra Maier-Salamon et al.
Annals of the New York Academy of Sciences, 1290, 98-106 (2013-07-17)
Resveratrol exhibits a variety of biological and pharmacological activities despite its extensive metabolism to sulfates and glucuronides in the intestine and liver. The metabolism of resveratrol is cell specific and strongly correlates with enzyme expression levels. However, a high rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service