Skip to Content
Merck
All Photos(1)

Key Documents

76130

Supelco

Ethyl acrylate

analytical standard

Synonym(s):

Acrylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOC2H5
CAS Number:
Molecular Weight:
100.12
Beilstein:
773866
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

vapor pressure

31 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

721 °F

shelf life

limited shelf life, expiry date on the label

contains

~0.002% hydroquinone monomethyl ether as stabilizer

expl. lim.

12.1 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.406 (lit.)
n20/D 1.406

bp

99 °C (lit.)

mp

−71 °C (lit.)

density

0.921 g/mL at 20 °C
0.918 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
petroleum

format

neat

SMILES string

CCOC(=O)C=C

InChI

1S/C5H8O2/c1-3-5(6)7-4-2/h3H,1,4H2,2H3

InChI key

JIGUQPWFLRLWPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Ethyl acrylate can undergo anionic polymerization upon initiation by tetrabutylammonium azide in the presence of alkylaluminum bisphenoxides in toluene to yield azide-end poly(ethyl acrylate) (PEA).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anionic polymerization of ethyl acrylate initiated by tetrabutylammonium azide: direct synthesis of end-clickable polyacrylate
Kataoka Y, et al.
Polym. Chem. (2017)
Marilene S Oliveira et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 10(10), 1546-1555 (2011-07-08)
Singlet molecular oxygen O(2)((1)Δ(g)) is a potent oxidant that can react with different biomolecules, including DNA, lipids and proteins. Many polycyclic aromatic hydrocarbons have been studied as O(2)((1)Δ(g)) chemical traps. Nevertheless, a suitable modification in the polycyclic aromatic ring must
Jiaan Shao et al.
Organic letters, 14(21), 5452-5455 (2012-10-26)
Palladium-catalyzed C-H functionalization using guanidine as the directing group was achieved under mild reaction conditions. Various guanidine derivatives were produced in moderate to good yields by using simple unactivated arenes or ethyl acrylate as the source of arylation or olefination
J C O Villanova et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 79(3), 664-673 (2011-09-29)
The design of new excipients that extend the release of drugs from tablets over prolonged periods is essential in reaching enhanced therapeutic performances. In this sense, the objective of this study was to develop new excipients, based on acrylic monomers
Anna E V Hagström et al.
Biotechnology and bioengineering, 102(3), 693-699 (2008-09-30)
An OH-functional polyester has been acrylated via transesterification of ethyl acrylate, catalyzed by Candida antarctica lipase B (CalB) in two different preparations: Novozym 435 and immobilized on Accurel MP1000. The batch process resulted in incomplete acrylation as well as severe

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service