Skip to Content
Merck
All Photos(1)

Key Documents

65990

Sigma-Aldrich

(±)-2-Methyl-1-butanol

≥98.0% (GC)

Synonym(s):

sec-Butyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1839383
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

refractive index

n20/D 1.411

bp

126-129 °C (lit.)

density

0.819 g/mL at 20 °C (lit.)

SMILES string

CCC(C)CO

InChI

1S/C5H12O/c1-3-5(2)4-6/h5-6H,3-4H2,1-2H3

InChI key

QPRQEDXDYOZYLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

108.5 °F - closed cup

Flash Point(C)

42.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viviane Cordovez et al.
Frontiers in plant science, 8, 1262-1262 (2017-08-09)
Beneficial soil microorganisms can affect plant growth and resistance by the production of volatile organic compounds (VOCs). Yet, little is known on how VOCs from soil-borne plant pathogens affect plant growth and resistance. Here we show that VOCs released from
Lawrence M Hanks et al.
Journal of economic entomology, 112(5), 2236-2252 (2019-05-29)
We present research on the chemical ecology of 14 species of longhorned beetles (Coleoptera: Cerambycidae), in four tribes of the subfamily Cerambycinae, conducted in east-central Illinois over 8 yr. Adult males produce aggregation-sex pheromones that attract both sexes. Twenty independent
Hyung Min Kim et al.
Journal of AOAC International, 100(5), 1492-1499 (2017-04-20)
A simple and fast method was developed for the determination of volatile organic compounds in alcoholic beverages. Eleven volatile organic compounds (acetaldehyde, methanol, 2-propanol, tert-butanol, 1-propanol, ethyl acetate, 2-butanol, isobutanol, 1-butanol, 3-methyl-1butanol, and 2-methyl-1-butanol) in alcoholic beverages were analyzed with
Marianthi Karali et al.
PloS one, 6(7), e22166-e22166 (2011-08-06)
Gene transfer using adeno-associated viral (AAV) vectors has been successfully applied in the retina for the treatment of inherited retinal dystrophies. Recently, microRNAs have been exploited to fine-tune transgene expression improving therapeutic outcomes. Here we evaluated the ability of retinal-expressed
Eduardo Coelho et al.
Food chemistry, 323, 126826-126826 (2020-04-27)
During beer ageing, endogenous barrel microbes grow spontaneously and transform wort/beer composition, being Dekkera bruxellensis and Saccharomyces cerevisiae among the main contributors to the chemical and sensory profile of aged beer. This work aims at the application of multi-starter cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service