Skip to Content
Merck
All Photos(1)

Key Documents

60360

Sigma-Aldrich

Potassium phthalate monobasic

puriss. p.a., ≥99.5% (T)

Synonym(s):

KHP, Phthalic acid monopotassium salt, Potassium biphthalate, Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCC6H4COOK
CAS Number:
Molecular Weight:
204.22
Beilstein:
3637128
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Quality Level

Assay

≥99.5% (T)

form

solid

pH

4

mp

295-300 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium phthalate monobasic (KHP) is the monopotassium salt of pthalic acid. It has been proposed as a certified reference material (CRM) under ISO/IEC 17025 and ISO Guide 34 accreditation. Its spectral shift, multiplicity, solubility and relaxation times in different solvents have been reported.

Application

Potassium phthalate monobasic may be used as a primary standard in acidimetric titration and also to prepare potassium phthalate buffer solution for pH calibration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Padró et al.
Talanta, 107, 95-102 (2013-04-20)
Dispersive ionic liquid-liquid microextraction combined with liquid chromatography and UV detection was used for the determination of two antichagasic drugs in human plasma: nifurtimox and benznidazole. The effects of experimental parameters on extraction efficiency-the type and volume of ionic liquid
2, 4, 6-Trinitrobenzoic acid as a primary standerd in acidimetry.
Smith GF and Wilkins DH.
Analytica Chimica Acta, 8, 209-216 (1953)
Determination of standard pH values for potassium hydrogen phthalate reference buffer solutions in 10, 20, 50, 64 and 84.2 wt per cent methanol/water mixed solvents at temperatures from 283.15 to 313.15 K.
Mussini T, et al.
Electrochimica Acta, 28(11), 1593-1598 (1983)
Maria Cristina Fossi et al.
Marine pollution bulletin, 64(11), 2374-2379 (2012-09-12)
Baleen whales are potentially exposed to micro-litter ingestion as a result of their filter-feeding activity. However, the impacts of microplastics on baleen whales are largely unknown. In this case study of the Mediterranean fin whale (Balaenoptera physalus), we explore the
Randi J Bertelsen et al.
Environmental health perspectives, 121(2), 251-256 (2012-11-21)
High-molecular-weight phthalates in indoor dust have been associated with asthma in children, but few studies have evaluated phthalate biomarkers in association with respiratory outcomes. We explored the association between urinary concentrations of phthalate metabolites and current asthma. In a cross-sectional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service