Skip to Content
Merck
All Photos(1)

Key Documents

53436

Millipore

Perfringens S.F.P. Selective Supplement

suitable for microbiology

Synonym(s):

Antibiotic supplement, S.F.P. Selective Supplement, Shahidi Ferguson Perfringens Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 5 vials

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Clostridium spp.

General description

Perfringens S.F.P. Selective Supplement is used as an antibiotic supplement used in culture media to inhibit the growth of unwanted organisms while allowing the growth of the desired microorganisms. This antibiotic supplement is recommended for the selective isolation and enumeration of Clostridium perfringens. The supplement consists of a combination of kanamycin sulfate, an aminoglycoside antibiotic derived from Streptomyces kanamyceticus, and polymyxin B, an antibiotic effective against resistant gram-negative bacteria, derived from Bacillus polymyxa. Polymyxin B exhibits bactericidal action against most gram-negative bacilli, excluding the Proteus group, thereby selectively allowing the growth of Clostridium perfringens.

Application

Perfringens S.F.P. Selective Supplement is recommended for the selective isolation and enumeration of Clostridium perfringens from various food and clinical samples.

Components

(per vial, sufficient for 500 ml medium):
Kanamycin sulphate 6 mg
Polymyxin B 15000 IU

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service