Skip to Content
Merck
All Photos(2)

Documents

50409

Supelco

Thymol

Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[(CH3)2CH]C6H3-5-(CH3)OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1907135
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

Standard for quantitative NMR
certified reference material

Quality Level

vapor pressure

1 mmHg ( 64 °C)

description

qNMR Standard

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

LC/MS: suitable
qNMR: suitable

bp

232 °C (lit.)

mp

48-51 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
pharmaceutical

format

neat

SMILES string

CC(C)c1ccc(C)cc1O

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3

InChI key

MGSRCZKZVOBKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out our entire range of quantitative NMR standards (qNMR standards)
Thymol, a phenolic monoterpene derivative of cymene and an isomer of carvacrol, is a common constituent of the essential oils, derived from Thymus and Origanum plants. It is a flavoring compound and its content is directly proportional to the characteristic flavor and quality of thyme and ajwain essential oils. Thymol finds a variety of applications in pharmaceuticals, cosmetics, and the food industry.

Application

Thymol has been used as certified reference material for the quantification of the analyte in essential oil from the leaves of Mentha longifolia Huds. (Lamiaceae) and Mentha spicata L. (Lamiaceae) using gas chromatography coupled to mass spectrometry (GC-MS).

Other Notes

This compound is commonly found in plants of the genus: ginkgo thymus

Recommended products

Check out ChemisTwin, our brand new online portal for identity confirmation of NMR spectra. A feature for quantitative NMR is in development. Learn more or reach out to us for a free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical characterization, antimicrobial and antioxidant properties of Mentha spicata L.(Lamiaceae) essential oil
Niksic H, et al.
Bulletin of the Chemists and Technologists of Bosnia and Herzegovina, 43-48 (2018)
Seasonal variation in content and chemical composition of essential oils from leaves of Mentha longifolia Huds.(Lamiaceae)
Niksic H, et al.
Bulletin of the Chemists and Technologists of Bosnia and Herzegovina, 43, 29-34 (2014)
J D Evans et al.
Current microbiology, 41(5), 336-340 (2000-10-03)
Thymol (5-methyl-2-isopropylphenol) is a phenolic compound that is used to inhibit oral bacteria. Because little is known regarding the effects of this compound on ruminal microorganisms, the objective of this study was to determine the effects of thymol on growth
Production of thymol rich extracts from ajwain (Carum copticum L.) and thyme (Thymus vulgaris L.) using supercritical CO2
Morsy NFS
Industrial Crops and Products, 145(5), 112072-112072 (2020)
Thymol, thyme, and other plant sources: Health and potential uses
Salehi B, et al.
Phytotherapy Research, 32(9), 1688-1706 (2018)

Articles

Learn about ChemisTwin® Portal, an online analytical spectra interpretation tool. Discover its NMR lab benefits, workflow integration, and user responsibilities. Explore dRMs, algorithm accuracy, data formats, subscription possibilities, and more.

Learn about ChemisTwin® Portal, an online analytical spectra interpretation tool. Discover its NMR lab benefits, workflow integration, and user responsibilities. Explore dRMs, algorithm accuracy, data formats, subscription possibilities, and more.

Learn about ChemisTwin® Portal, an online analytical spectra interpretation tool. Discover its NMR lab benefits, workflow integration, and user responsibilities. Explore dRMs, algorithm accuracy, data formats, subscription possibilities, and more.

Learn about ChemisTwin® Portal, an online analytical spectra interpretation tool. Discover its NMR lab benefits, workflow integration, and user responsibilities. Explore dRMs, algorithm accuracy, data formats, subscription possibilities, and more.

Related Content

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

ChemisTwin automates NMR result interpretation, saving time with structure confirmation and quantitative NMR measurements using electronic Reference Materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service