Skip to Content
Merck
All Photos(1)

Documents

46432

Supelco

Metsulfuron-methyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N5O6S
CAS Number:
Molecular Weight:
381.36
Beilstein:
587472
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2

InChI

1S/C14H15N5O6S/c1-8-15-12(18-14(16-8)25-3)17-13(21)19-26(22,23)10-7-5-4-6-9(10)11(20)24-2/h4-7H,1-3H3,(H2,15,16,17,18,19,21)

InChI key

RSMUVYRMZCOLBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metsulfuron-methyl belongs to the class of post-emergent, low-application-rate sulfonylurea herbicides. It is used in cereals and other row crops for protection against broad-leaved weeds and some grasses.

Application

Metsulfuron-methyl may be used as an analytical reference standard for the determination of the analyte in water samples and environmental samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination and quantitation of ten sulfonylurea herbicides in soil samples using liquid chromatography with electrospray ionization mass spectrometric detection.
Guibiao Y, et al.
Chinese Journal of Analytical Chemistry, 34(2-3), 1207-1212 (2006)
Persistence of metsulfuron-methyl in wheat crop and soil.
Sondhia S
Environmental Monitoring and Assessment, 147(1-3), 463-469 (2008)
Preconcentration and determination of nicosulfuron, thifensulfuron-methyl and metsulfuron-methyl in water samples using carbon nanotubes packed cartridge in combination with high performance liquid chromatography.
Zhou Q, et al.
Analytica Chimica Acta, 559(2), 200-206 (2006)
Qiang Tang et al.
Journal of environmental sciences (China), 24(8), 1492-1497 (2012-01-01)
Tribenuron methyl (TBM) is a member of the sulfonylurea herbicide family and is widely used worldwide. In this study, TBM-degrading bacteria were enriched with TBM as potential carbon, nitrogen or sulfur source, and 44 bacterial isolates were obtained. These isolates
B S Ismail et al.
Journal of environmental biology, 33(3), 573-577 (2012-10-04)
Laboratory experiments were conducted to evaluate adsorption, desorption and mobility of metsulfuron-methyl in soils of the oil palm agroecosystem consisting of the Bernam, Selangor, Rengam and Bongor soil series. The lowest adsorption of metsulfuron-methyl occurred in the Bongor soil (0.366

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service