Skip to Content
Merck
All Photos(1)

Key Documents

45997

Supelco

Methyl acetate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH3
CAS Number:
Molecular Weight:
74.08
Beilstein:
1736662
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.55 (vs air)

vapor pressure

165 mmHg ( 20 °C)

Assay

≥99.9% (GC)

autoignition temp.

936 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

16 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.361 (lit.)
n20/D 1.362

bp

57-58 °C (lit.)

mp

−98 °C (lit.)

density

0.934 g/mL at 25 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COC(C)=O

InChI

1S/C3H6O2/c1-3(4)5-2/h1-2H3

InChI key

KXKVLQRXCPHEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl acetate is an acetate ester, classified as a volatile organic compound (VOC). It is used as a solvent for many resins and oils.

Application

The analytical standard can also be used as follows:

  • Quantitative analysis of seven volatile organic compounds (VOCs) in paint coating samples by dynamic headspace-gas chromatography-mass spectrometry (D-HS-GC-MS)
  • Detection of 88 VOCs in breath samples of lung cancer patients using solid-phase microextraction (SPME) combined with GC-MS
  • Development of a thermal desorption-gas chromatography-mass spectrometry (TDS-GC/MS) based method for the identification of very volatile organic compounds (VVOCs) in indoor air samples

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Yang et al.
The journal of physical chemistry. A, 112(28), 6364-6372 (2008-06-21)
The mechanisms and the kinetics of the OH (OD) radicals with methyl acetate CH3C(O)OCH3 are investigated theoretically. The dual-level direct dynamics method is employed in the calculation of the rate constants. The optimized geometries and frequencies and the gradients of
Benjamin Bechem et al.
The Journal of organic chemistry, 75(5), 1795-1798 (2010-02-06)
5-Substituted-2-furan methanols 1a-c are subject to enantioselective carbonyl allylation, crotylation and tert-prenylation upon exposure to allyl acetate, alpha-methyl allyl acetate, or 1,1-dimethylallene in the presence of an ortho-cyclometalated iridium catalyst modified by (R)-Cl,MeO-BIPHEP, (R)-C3-TUNEPHOS, and (R)-C3-SEGPHOS, respectively. In the presence
Nicolas Salem et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 13(1), 140-151 (2010-04-20)
Studies have established the value of [(methyl)1-(11)C]-acetate ([(11)C]Act) combined with 2-deoxy-2[(18)F]fluoro-D-glucose (FDG) for detecting hepatocellular carcinoma (HCC) using positron emission tomography (PET). In this study, the metabolic fate of [(11)C]Act in HCC was characterized. Experiments with acetic acid [1-(14)C] sodium
Saman Mohammadi et al.
PloS one, 9(9), e106653-e106653 (2014-09-11)
In this study, we compared, for the first time, the release of a 432 kDa prostaglandin F2a analogue drug, Latanoprost, from commercially available contact lenses using in vitro models with corneal epithelial cells. Conventional polyHEMA-based and silicone hydrogel soft contact
Franc Avbelj et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(9), 3137-3141 (2009-02-10)
Recent calorimetric measurements of the solvation enthalpies of some dipeptide analogs confirm our earlier prediction that the principle of group additivity is not valid for the interaction of the peptide group with water. We examine the consequences for understanding the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service