Skip to Content
Merck
All Photos(1)

Documents

45429

Supelco

Dibrom®

PESTANAL®, analytical standard

Synonym(s):

(1,2-Dibromo-2,2-dichloroethyl) dimethyl phosphate, Bromchlophos, Naled

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7Br2Cl2O4P
CAS Number:
Molecular Weight:
380.78
Beilstein:
2049930
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=O)(OC)OC(Br)C(Cl)(Cl)Br

InChI

1S/C4H7Br2Cl2O4P/c1-10-13(9,11-2)12-3(5)4(6,7)8/h3H,1-2H3

InChI key

BUYMVQAILCEWRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Dibrom is a registered trademark of AMVAC Chemical Corporation
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Tham C Hoang et al.
Environmental toxicology and chemistry, 30(4), 997-1005 (2011-02-11)
Honeybees are the standard insect test species used for toxicity testing of pesticides on nontarget insects for the U.S. Environmental Protection Agency (U.S. EPA) under the Federal Insecticide Fungicide and Rodenticide Act (FIFRA). Butterflies are another important insect order and
Zandra Duprey et al.
Journal of the American Mosquito Control Association, 24(1), 42-46 (2008-04-29)
In October 2004, the Florida Department of Health (FLDOH) and the Centers for Disease Control and Prevention (CDC) assessed human exposure to ultra-low volume (ULV) aerial application of naled. Teams administered activity questionnaires regarding pesticide exposure and obtained baseline urine
H Zhong et al.
Environmental entomology, 39(6), 1961-1972 (2011-12-21)
We assessed the exposure and acute toxicity of naled, applied aerially as an ultra-low-volume spray for mosquito control, on late instar larvae of the Miami blue (Cyclargus thomasi bethunebakeri) (Comstock and Huntington 1943) (Lepidoptera: Lycaenidae), an imperiled South Florida butterfly.
Jerome J Schleier et al.
Archives of environmental contamination and toxicology, 58(1), 105-111 (2009-06-19)
One of the most effective ways of managing adult mosquitoes that vector human and animal pathogens is the use of ultra-low-volume (ULV) insecticides. Because of the lack of environmental fate studies and concerns about the safety of the insecticides used
Timothy A Bargar
Environmental toxicology and chemistry, 31(9), 2124-2129 (2012-06-29)
The relationship between total cholinesterase activity (TChE) and mortality in four butterfly species (great southern white [Ascia monuste], common buckeye [Junonia coenia], painted lady [Vanessa cardui], and julia butterflies [Dryas julia]) was investigated. Acute contact toxicity studies were conducted to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service