Skip to Content
Merck
All Photos(1)

Key Documents

43987

Supelco

(S)-Mevalonic acid lithium salt

analytical standard

Synonym(s):

Lithium (S)-3,5-dihydroxy-3-methyl-pentanoate, Lithium (S)-3,5-dihydroxy-3-methyl-valerate, S-MVA-Li

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O4 · xLi+
CAS Number:
Molecular Weight:
148.16 (free acid basis)
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

InChI

1S/C6H12O4/c1-6(10,2-3-7)4-5(8)9/h7,10H,2-4H2,1H3,(H,8,9)/t6-/m0/s1

InChI key

KJTLQQUUPVSXIM-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite of the mevalonate pathway, which plays a key role in the biosynthesis of sterols, dolichol, heme and ubiquinone. Of interest for research in the disease areas oncology, autoimmune diseases, artherosclerosis and Alzheimer disease, as well as for inherited deficiencies of mevalonate kinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service