Skip to Content
Merck
All Photos(1)

Key Documents

36551

Supelco

Chlorfenvinphos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14Cl3O4P
CAS Number:
Molecular Weight:
359.57
Beilstein:
8712900
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of Z and E isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)O\C(=C\Cl)c1ccc(Cl)cc1Cl

InChI

1S/C12H14Cl3O4P/c1-3-17-20(16,18-4-2)19-12(8-13)10-6-5-9(14)7-11(10)15/h5-8H,3-4H2,1-2H3/b12-8-

InChI key

FSAVDKDHPDSCTO-WQLSENKSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorfenvinphos is an organophosphorus insecticide used for the control of the bee parasite Varroa destructor and also found to be effective against a variety of insects affecting vegetables, fruits and crops.

Application

Chlorfenvinphos may be used as an analytical reference standard for the quantification of the analyte in bee-wax, grains and environmental samples using different analytical techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Paul Trethowan et al.
Ecology and evolution, 7(1), 253-262 (2017-01-11)
It has been proposed that there is a thermal cost of the mane to male lions, potentially leading to increased body surface temperatures (
Differential pulse polarographic determination of dicrotophos, crotoxyphos and chlorfenvinphos in grains and soils
Sreedhar Y.N, et al.
Talanta, 44(10), 1859-1863 (1997)
Determination of amitraz and other acaricide residues in beeswax.
Korta E, et al.
Analytica Chimica Acta, 475(1-2), 97-103 (2003)
Marta O Barbosa et al.
Analytical and bioanalytical chemistry, 408(29), 8355-8367 (2016-10-14)
Organic micropollutants present in drinking water (DW) may cause adverse effects for public health, and so reliable analytical methods are required to detect these pollutants at trace levels in DW. This work describes the first green analytical methodology for multi-class
Juan L Acero et al.
Journal of hazardous materials, 153(1-2), 320-328 (2007-10-02)
Unknown second-order rate constants for the reactions of three organophosphorus pesticides (chlorpyrifos, chlorfenvinfos and diazinon) with chlorine were determined in the present study, and the influence of pH and temperature was established. It was found that an increase in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service