Skip to Content
Merck
All Photos(1)

Key Documents

36151

Supelco

Mepiquat chloride

PESTANAL®, analytical standard

Synonym(s):

1,1-Dimethylpiperidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16ClN
CAS Number:
Molecular Weight:
149.66
Beilstein:
3914089
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Cl-].C[N+]1(C)CCCCC1

InChI

1S/C7H16N.ClH/c1-8(2)6-4-3-5-7-8;/h3-7H2,1-2H3;1H/q+1;/p-1

InChI key

VHOVSQVSAAQANU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Mepiquat chloride is a plant growth regulator, which is basically employed in cotton cultivation, in order to control the growth rate and hence increase the quality and yield.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Managing mepiquat chloride and plant density for optimal yield and quality of cotton
Ren X, et al.
Field Crops Research, 149, 1-10 (2013)
Jay A Rosenheim
Journal of economic entomology, 106(3), 1286-1293 (2013-07-20)
Because the farmer is typically excluded from the experimental research setting, experimental research may face challenges in evaluating pest management tactics whose costs and benefits hinge on farmer decision-making. In these cases an ecoinformatics approach, in which observational data collected
Sonja Riediker et al.
Journal of chromatography. A, 966(1-2), 15-23 (2002-09-07)
A sensitive and selective method is presented for the simultaneous analysis of the pesticides chlormequat and mepiquat at trace levels in tomato, pear, and wheat flour. The method entails direct injection of the food extract onto an on-line solid-phase extraction
Addissu G Ayele et al.
Plants (Basel, Switzerland), 9(7) (2020-07-11)
Verticillium wilt, caused by Verticillium dahliae Kleb., leads to significant losses in cotton yield and fiber quality worldwide. To investigate Verticillium wilt impact on photosynthesis rate, yield, and fiber quality, six upland cotton genotypes, namely Verticillium susceptible (DP 1612 B2XF)
Wen-Xi Li et al.
Ecotoxicology and environmental safety, 85, 137-143 (2012-09-08)
A simple, accurate and highly sensitive analytical method was developed in this study for determining the residues and dissipation dynamics of mepiquat chloride in soil and cotton crops (including plant and seed). The samples in this method were directly extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service