Skip to Content
Merck
All Photos(2)

Documents

34230

Supelco

(E)-Metominostrobin

PESTANAL®, analytical standard

Synonym(s):

Methyl (E)-α-methoxyimino-N-methyl-2-phenoxyphenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2O3
CAS Number:
Molecular Weight:
284.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)C(=N\OC)\c1ccccc1Oc2ccccc2

InChI

1S/C16H16N2O3/c1-17-16(19)15(18-20-2)13-10-6-7-11-14(13)21-12-8-4-3-5-9-12/h3-11H,1-2H3,(H,17,19)/b18-15+

InChI key

HIIRDDUVRXCDBN-OBGWFSINSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metominostrobin is a systemic fungicide mainly used against rice blast disease. Its mode of action involves inhibiting respiratory electron transfer.

Application

(E)-Metominostrobin may be used as an analytical reference standard for the determination of the analyte in agricultural products, food matrices, water and fruit samples by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and Easy Multiresidue Method for Determination of Pesticide Residues in Foods Using Gas or Liquid Chromatography-Tandem Mass Spectrometry.
Pesticides: Strategies for Pesticides Analysis (2011)
Current status of conventional and molecular interventions for blast resistance in rice.
Srivastava D, et al.
Rice Science, 24(6), 299-321 (2017)
Combination of solvent extractants for dispersive liquid-liquid microextraction of fungicides from water and fruit samples by liquid chromatography with tandem mass spectrometry.
Pastor-Belda M, et al.
Food Chemistry, 233(5), 69-76 (2017)
Validation of multiresidue screening methods for the determination of 186 pesticides in 11 agricultural products using gas chromatography (GC).
Hirahara Y, et al.
Journal of Health Science, 51(5), 617-627 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service