Skip to Content
Merck
All Photos(1)

Key Documents

34093

Supelco

Closantel

PESTANAL®, analytical standard

Synonym(s):

5′-Chloro-4′-(4-chloro-α-cyanobenzyl)-3,5-diiodo-2′-methylsalicylanilide, N-[5-Chloro-4-(4-chloro-α-cyanobenzyl)-2-methylphenyl]-2-hydroxy-3,5-diiodobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14Cl2I2N2O2
CAS Number:
Molecular Weight:
663.07
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(C(C#N)c2ccc(Cl)cc2)c(Cl)cc1NC(=O)c3cc(I)cc(I)c3O

InChI

1S/C22H14Cl2I2N2O2/c1-11-6-15(17(10-27)12-2-4-13(23)5-3-12)18(24)9-20(11)28-22(30)16-7-14(25)8-19(26)21(16)29/h2-9,17,29H,1H3,(H,28,30)

InChI key

JMPFSEBWVLAJKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Closantel belongs to the salicylanilide class of anthelmintics, that can extensively bind to the plasma albumin. It can be effectively used against, a vast range of blood feeding internal parasites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Closantel toxicosis in kid goats.
R Ecco et al.
The Veterinary record, 159(17), 564-566 (2006-10-24)
Yumi Kumagai et al.
Infection and immunity, 74(9), 5014-5022 (2006-08-24)
Ehrlichia chaffeensis, the etiologic agent of human monocytic ehrlichiosis, replicates in early endosomes by avoiding lysosomal fusion in monocytes and macrophages. In E. chaffeensis we predicted three pairs of putative two-component regulatory systems (TCSs) designated PleC-PleD, NtrY-NtrX, and CckA-CtrA based
Quantitative determination of Closantel residues in milk by high-performance liquid chromatography with fluorescence detection
Stove.G, et al.
Journal of Chromatography A, 846, 383-386 (1999)
K M Cooper et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(8), 1263-1271 (2012-05-29)
The ProSafeBeef project studied the prevalence of residues of anthelmintic drugs used to control parasitic worms and fluke in beef cattle in Ireland. Injured (casualty) cattle may enter the human food chain under certain conditions, verified by an attending veterinarian
Haemonchus contortus: the uptake and metabolism of closantel
Rothwell J and Sangster N
International Journal For Parasitology, 27, 313-319 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service