Skip to Content
Merck
All Photos(1)

Documents

31635

Supelco

Fenazaquin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O
CAS Number:
Molecular Weight:
306.40
Beilstein:
8331263
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(C)c1ccc(CCOc2ncnc3ccccc23)cc1

InChI

1S/C20H22N2O/c1-20(2,3)16-10-8-15(9-11-16)12-13-23-19-17-6-4-5-7-18(17)21-14-22-19/h4-11,14H,12-13H2,1-3H3

InChI key

DMYHGDXADUDKCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Houneïda Benbouzid et al.
Communications in agricultural and applied biological sciences, 74(1), 129-135 (2009-01-01)
Our biodiversity has long been preserved, but the main constituents of our environment have been particularly affected by the addition of molecules resulting from agricultural and industrial activities. It is well accepted that these changes may stress some species, making
Anil Duhan et al.
Bulletin of environmental contamination and toxicology, 84(2), 217-220 (2009-10-23)
Fenazaquin (4-[[4 (1,1-dimethylethyl) phenyl] ethoxy]quinazoline) is a new acaricide of the quinazoline class. Residue levels of fenazaquin were determined in unprocessed and processed okra fruits to evaluate the effect of different processes (washing, boiling and washing followed by boiling) in
Metabolism of fenazaquin, an acaricide in tea plant.
J Bhattacharyya et al.
Bulletin of environmental contamination and toxicology, 75(3), 569-573 (2006-01-03)
Vipin Kumar et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 44(4), 596-600 (2006-04-26)
Fenazaquin is a non-systemic acaricide/insecticide used widely in controlling mites and other related pests in fruits, vegetables and tea. The objective of this research was to investigate the disappearance trend in tea of fenazaquin residue level and its transfer in
N V Kirby et al.
Pest management science, 57(9), 844-851 (2001-09-20)
A series of novel (hetero) aryloxylepidine derivatives was devised as hybrid structures of the phenoxyquinoline and phenethoxyquin(az)oline fungicides. Synthesis of these targets required the development of several new routes to derivatised 4-hydroxymethylquinolines, and subsequent coupling with phenols or haloarenes. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service