Skip to Content
Merck
All Photos(1)

Documents

17935

Supelco

Hydrogen chloride solution

~1.25 M in methanol, for GC derivatization, LiChropur

Synonym(s):

Hydrochloric acid in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCl · CH3OH
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

product name

Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur

grade

for GC derivatization

Quality Level

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

concentration

~1.25 m HCl (T)

technique(s)

gas chromatography (GC): suitable

density

0.831 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Esterification of galacturonic acid and polyuronides with methanol-hydrogen chloride.
E F JANSEN et al.
Journal of the American Chemical Society, 68, 1475-1477 (1946-08-01)
A REACTION TUBE FOR METHANOLYSIS; INSTABILITY OF HYDROGEN CHLORIDE IN METHANOL.
Y KISHIMOTO et al.
Journal of lipid research, 6, 435-436 (1965-07-01)
Liquid chromatography--high resolution mass spectrometry analysis of fatty acid metabolism
Kamphorst JJ, et al.
Analytical Chemistry, 83(23), 9114-9122 (2011)
Michael F. L'Annunziata
Handbook of Radioactivity Analysis, 400-400 (2012)
Jay G Hosking et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(4), 1005-1015 (2014-10-21)
Successful decision making often requires weighing a given option's costs against its associated benefits, an ability that appears perturbed in virtually every severe mental illness. Animal models of such cost/benefit decision making overwhelmingly implicate mesolimbic dopamine in our willingness to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service