Skip to Content
Merck
All Photos(1)

Documents

02343

Supelco

Octocrylene

analytical standard

Synonym(s):

2-Ethylhexyl 2-cyano-3,3-diphenylacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2C=C(CN)CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
361.48
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.567 (lit.)
n20/D 1.567±0.001

bp

218 °C/1.5 mmHg (lit.)

mp

−10 °C (lit.)

density

1.051 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCC(CC)COC(=O)\C(C#N)=C(/c1ccccc1)c2ccccc2

InChI

1S/C24H27NO2/c1-3-5-12-19(4-2)18-27-24(26)22(17-25)23(20-13-8-6-9-14-20)21-15-10-7-11-16-21/h6-11,13-16,19H,3-5,12,18H2,1-2H3

InChI key

FMJSMJQBSVNSBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octocrylene is a commercially used UVB filter, termed as an environmental pollutant because of its stability, lipophilic nature, and resistance to sunlight degradation. It is employed as an ingredient in water resistant sunscreen products and high SPF formulations.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Octocrylene may be used as an analytical reference standard for the quantification of the analyte in the following:        
  • Sunscreen formulations using reversed-phase high-performance liquid chromatography (RP-HPLC) technique.   
  • Marine mammals using ultra high-performance liquid chromatography (UHPLC).       
  • Solid lipid nanoparticle systems using high-performance liquid chromatography (HPLC) technique.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

453.2 °F - closed cup

Flash Point(C)

234 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

First determination of UV filters in marine mammals. Octocrylene levels in Franciscana dolphins
Gago-Ferrero P, et al.
Environmental Science & Technology, 47(11), 5619-5625 (2013)
Analytical method development for simultaneous estimation of Oxybenzone, Octocrylene, Octinoxate and Avobenzone in sunscreen by high performance liquid chromatography and its validation
Bhuva C, et al.
Pharmacophore, 3(2) (2012)
Rosario Rodil et al.
Journal of chromatography. A, 1179(2), 81-88 (2007-12-22)
Stir bar sorptive extraction (SBSE) in combination with thermal desorption-gas chromatography-mass spectrometry (TD-GC-MS) was applied for the determination of 9 UV filters in water samples. The stir bar coated with polydimethylsiloxane (PDMS) was added to 20 mL of water sample
M S Berkman et al.
Die Pharmazie, 66(2), 105-110 (2011-03-26)
UV filters are traditionally classified as chemical absorbers and physical blockers depending on their mechanism of action. In this study, one of the most important chemical UVB absorber, octocrylene, was incorporated into Solid Lipid Nanoparticle (SLN) systems which themselves have
N Negreira et al.
Analytical and bioanalytical chemistry, 400(2), 603-611 (2011-02-22)
This work describes an effective, low solvent consumption and affordable sample preparation approach for the determination of eight UV filters in surface and wastewater samples. It involves sorptive extraction of target analytes in a disposable, technical grade silicone disc (5 mm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service