Skip to Content
Merck
All Photos(1)

Key Documents

24-4840

Sigma-Aldrich

Potassium iodate solution

1/60 M

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KIO3
CAS Number:
Molecular Weight:
214.00
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

1/10 N
1/60 M

SMILES string

[K+].[O-]I(=O)=O

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabian Rohner et al.
Food and nutrition bulletin, 33(4 Suppl), S330-S335 (2013-03-01)
Despite considerable progress made in the past decade through salt iodization programs, over 2 billion people worldwide still have inadequate iodine intake, with devastating consequences for brain development and intellectual capacity. To optimize these programs with regard to salt iodine
Ralf Biebinger et al.
The British journal of nutrition, 102(9), 1362-1369 (2009-08-06)
Adverse sensory changes prevent the addition of highly bioavailable ferrous sulfate (FeSO4) to most wheat flours. Poorly absorbable reduced Fe powders are commonly used. Encapsulation of FeSO4 can overcome these sensory changes, but the particle size of commercial compounds is
Mohsen Zeeb et al.
Talanta, 78(2), 584-589 (2009-02-11)
In this work, an on-line system with vapor-phase generation (VPG) and Fourier transform infrared (FTIR) spectrometric detection has been developed as a direct and highly selective analytical technique for the assay of penicillamine (PA). Potassium iodate solution was injected into
Tatsuya Kawai et al.
Academic radiology, 18(6), 682-689 (2011-03-12)
Evaluation of contrast enhancement of pulmonary lesions with ground-glass attenuation (GGA) is difficult with conventional techniques but might be possible using contrast-mapping images (CMIs) obtained by dual-energy computed tomography. To address this issue, a phantom study was conducted, and this
C Dryden et al.
Journal of perinatology : official journal of the California Perinatal Association, 27(5), 320-322 (2007-04-25)
Congenital thyrotoxicosis is a rare and potentially fatal illness. We report a case in a preterm infant delivered to a mother known to have autoimmune endocrine disease. Diagnosis was difficult because the infant's presenting symptoms and signs closely resembled congenital

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service