Skip to Content
Merck
All Photos(1)

Key Documents

24-1090

Sigma-Aldrich

Periodic acid

SAJ special grade, ≥98.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H5IO6
CAS Number:
Molecular Weight:
227.94
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ special grade

Assay

≥98.5%

form

crystalline

reaction suitability

reagent type: oxidant

availability

available only in Japan

mp

122 °C (lit.)

solubility

water: soluble

SMILES string

OI(O)(O)(O)(O)=O

InChI

1S/H5IO6/c2-1(3,4,5,6)7/h(H5,2,3,4,5,6,7)

InChI key

TWLXDPFBEPBAQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1B - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Murad Alturkustani et al.
Journal of neuropathology and experimental neurology, 74(3), 233-240 (2015-02-11)
The pathologic features of adult-onset leukoencephalopathy/leukodystrophy with axonal spheroids (ALAS) are variable, and this has led to different hypotheses as to whether primarily demyelination or axonopathy may underlie this disorder. Typical ALAS pathology is rarely accompanied by focal multiple sclerosis
Francesca Polverino et al.
The American journal of pathology, 185(3), 741-755 (2014-12-30)
Small animal models of chronic obstructive pulmonary disease (COPD) have several limitations for identifying new therapeutic targets and biomarkers for human COPD. These include a pulmonary anatomy that differs from humans, the limited airway pathologies and lymphoid aggregates that develop
Aranzazu Villasante et al.
Biomaterials, 35(22), 5785-5794 (2014-04-22)
Monolayer cultures of tumor cells and animal studies have tremendously advanced our understanding of cancer biology. However, we often lack animal models for human tumors, and cultured lines of human cells quickly lose their cancer signatures. In recent years, simple
Dmitriy Berillo et al.
Macromolecular bioscience, 12(8), 1090-1099 (2012-06-08)
Macroporous scaffolds composed of chitosan and using oxidized dextran as a crosslinker are produced through cryogelation. Introducing gelatin as a third component into the structure results in the formation of mesopores in the pore walls, which are not seen if
D Canbaz et al.
Allergy, 70(5), 522-532 (2015-02-14)
The initial immune response to house dust mite (HDM) is orchestrated by an interplay between epithelial cells (ECs) and dendritic cells (DCs). Innate cytokines released by HDM-exposed ECs activate airway DCs and effector inflammatory cells, which together induce a HDM-specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service