Skip to Content
Merck
All Photos(1)

Key Documents

24-0410

Sigma-Aldrich

Paraffin wax

SAJ first grade, mp 58-60 °C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

SAJ first grade

form

solid

availability

available only in Japan

mp

58-60 °C

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

602.6 °F - Pensky-Martens closed cup

Flash Point(C)

317 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Yong Zhang et al.
Journal of hazardous materials, 243, 105-111 (2012-10-31)
In this study, nanoscale zero-valent iron (NZVI) particles were synthesized and used for the reductive dehalogenation of short chain chlorinated paraffins (SCCPs) in the laboratory. The results show that the dechlorination rate of chlorinated n-decane (CP(10)) by NZVI increased with
Banu Dilek et al.
Archives of physical medicine and rehabilitation, 94(4), 642-649 (2012-11-29)
To evaluate the efficacy of paraffin bath therapy on pain, function, and muscle strength in patients with hand osteoarthritis. Prospective single-blinded randomized controlled trial. Department of physical medicine and rehabilitation in a university hospital. Patients with bilateral hand osteoarthritis (N=56).
B Hilger et al.
Environmental pollution (Barking, Essex : 1987), 175, 16-21 (2013-01-15)
Levels and distribution of chlorinated paraffins were studied in randomly taken house dust samples (<63 μm) from the greater area of Munich, Germany. Quantification of short- (SCCPs, C(10)-C(13)) and medium-chain chlorinated paraffins (MCCPs, C(14)-C(16)) were performed using gas chromatography electron capture
Sharoni Jacobs
Cold Spring Harbor protocols, 2012(11), 1203-1209 (2012-11-03)
The Whole Genome Sampling Analysis (WGSA) assay in combination with Affymetrix GeneChip Mapping Arrays is used for copy number analysis of high-quality DNA samples (i.e., samples that have been collected from blood, fresh or frozen tissue, or cell lines). Formalin-fixed
Elisabeth Erlacher et al.
Environmental pollution (Barking, Essex : 1987), 174, 236-243 (2013-01-05)
Crude oil is a complex mixture of petroleum hydrocarbons (PHC) with distinct chemical, physical and toxicological properties relevant for contaminated site risk assessment. Ecotoxicological effects of crude oil distillation fractions on luminescent bacteria (Vibrio fischeri), earthworms (Dendrobaena hortensis) and invertebrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service