Skip to Content
Merck
All Photos(1)

Key Documents

24-0320

Sigma-Aldrich

Paraffin wax

SAJ first grade, mp 40-42 °C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

SAJ first grade

form

solid

availability

available only in Japan

mp

40-42 °C

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

602.6 °F - Pensky-Martens closed cup

Flash Point(C)

317 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laiguo Chen et al.
Journal of chromatography. A, 1274, 36-43 (2013-01-05)
Accurately quantifying short chain chlorinated paraffins (SCCPs) in soil samples with gas chromatograph coupled with electron capture negative ionization mass spectrometry (GC-ECNI-MS) is difficult because many other polychlorinated pollutants are present in the sample matrices. These pollutants (e.g., polychlorinated biphenyls
Lindy Rauscher et al.
Journal of hazardous materials, 241-242, 441-449 (2012-10-24)
Surface waters impacted by urban runoff in metropolitan areas are becoming increasingly contaminated with polycyclic aromatic hydrocarbons (PAHs). Slow-release oxidant candles (paraffin-KMnO(4)) are a relatively new technology being used to treat contaminated groundwater and could potentially be used to treat
Zhi-Yong Zhang et al.
Journal of hazardous materials, 243, 105-111 (2012-10-31)
In this study, nanoscale zero-valent iron (NZVI) particles were synthesized and used for the reductive dehalogenation of short chain chlorinated paraffins (SCCPs) in the laboratory. The results show that the dechlorination rate of chlorinated n-decane (CP(10)) by NZVI increased with
Banu Dilek et al.
Archives of physical medicine and rehabilitation, 94(4), 642-649 (2012-11-29)
To evaluate the efficacy of paraffin bath therapy on pain, function, and muscle strength in patients with hand osteoarthritis. Prospective single-blinded randomized controlled trial. Department of physical medicine and rehabilitation in a university hospital. Patients with bilateral hand osteoarthritis (N=56).
Xue-Tong Wang et al.
Environmental science and pollution research international, 20(7), 4742-4749 (2013-01-08)
Short-chain chlorinated paraffins (SCCPs) are extremely complex technical mixtures of polychlorinated n-alkanes with carbon chain lengths from C10 to C13 and chlorine content between 49 and 70%. SCCPs are under consideration for inclusion in the Stockholm Convention on persistent organic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service