Skip to Content
Merck
All Photos(1)

Key Documents

18-0870

Sigma-Aldrich

Lead standard solution

suitable for atomic absorption spectrometry, 1 mg/mL Pb, 1000 ppm Pb

Synonym(s):

Lead(II) nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
liquid

form

liquid

availability

available only in Japan

concentration

1 mg/mL Pb
1000 ppm Pb

suitability

suitable for atomic absorption spectrometry

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1 - STOT RE 2

Target Organs

Blood,Central nervous system,Immune system,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xuemei Li et al.
Journal of hazardous materials, 213-214, 55-61 (2012-02-24)
An endophytic fungus was tested in rice (Oryza sativa L.) exposed to four levels of lead (Pb) stress (0, 50, 100 and 200 μM) to assess effects on plant growth, photosynthesis and antioxidant enzyme activity. Under Pb stress conditions, endophyte-infected
Yujiao Zhu et al.
Fish & shellfish immunology, 34(2), 617-622 (2013-01-01)
Glucose regulated protein (GRP) located in endoplasmic reticulum (ER) was a member of heat shock protein (Hsp) family. The protective mechanism adapted to ER stimuli was closely related to GRP. GRP78, known as BiP, was one of central regulator responded
Kiyomitsu Nemoto et al.
The Journal of toxicological sciences, 38(1), 127-129 (2013-01-30)
We previously demonstrated the super-induced expression of the Grin2c gene encoding the N-methyl-D-aspartate receptor 2C subunit during the development of liver enlargement with hepatocellular hypertrophy induced by phenobarbital, clofibrate, or piperonyl butoxide. In the present study, we assessed whether or
Trivedi Mrugesh et al.
Acta poloniae pharmaceutica, 68(5), 653-656 (2011-09-21)
The present study was an attempt to investigate toxic effects of lead on red blood corpuscle (RBC) under in vitro conditions. Venous blood samples from adult, healthy, well-nourished male donors falling in age group of 25-30 years were collected in
Rami Alkhatib et al.
Plant cell reports, 30(8), 1427-1434 (2011-03-16)
Effect of various lead (Pb) concentrations on the systemic movement of RNA viruses was examined in tobacco plants. Prior to inoculation, plants were grown hydroponically for 6 days in Hoagland's solution supplemented with five concentrations of lead nitrate [Pb(NO(3))(2)]: 0.0 (control)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service