Skip to Content
Merck
All Photos(1)

Key Documents

11-0100

Sigma-Aldrich

Fehling′s reagent (B)

Synonym(s):

Potassium sodium tartrate solution, alkaline, Fehling’s reagent II for sugars

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

liquid

availability

available only in Japan

storage temp.

15-25°C

SMILES string

[Na+].[K+].OC(C(O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.K.Na/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);;/q;2*+1/p-2

InChI key

LJCNRYVRMXRIQR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service