Skip to Content
Merck
All Photos(1)

Key Documents

04634772001

Roche

Genopure Buffer Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.21

manufacturer/tradename

Roche

Quality Level

packaging

kit of for 20 maxi preps or 60 midi preps
for low copy number plasmids

General description

Doubling the volume of the Suspension, Lysis, and Neutralization Buffers during the Genopure plasmid preparation procedure will provide better yields of plasmid DNA from low-copy number plasmids. The buffers in this set may be used with either of the Genopure Plasmid Kits (Midi or Maxi).

Application

Doubling the volume of the Suspension, Lysis, and Neutralization Buffers during the Genopure plasmid preparation procedure will provide better yields of plasmid DNA from low-copy number plasmids. The buffers in this set may be used with either of the Genopure Plasmid Kits (Midi or Maxi).

Features and Benefits

The Genopure Buffer Set for Low-Copy Number Plasmids provides additional buffers to supplement those in the Genopure Plasmid Kits.

RNase A is also included in the set because it eliminates bacterial RNA from the preparation, thereby improving the recovery of low copy number plasmids.

  • Convenient, ready-to-use, function-tested, nuclease-free buffers
  • Helps the Genopure Plasmid Midi and Maxi Kits enhance plasmid yield
  • Ensures reproducible results

Components

  • Suspension Buffer, 500 mL
  • RNase A, 50 mg
  • Lysis Buffer, 500 mL
  • Neutralization Buffer, 500 mL

Other Notes

For life science research only. Not for use in diagnostic procedures.

To learn how these buffers are used, see the product listings or the package inserts for the Genopure Plasmid Midi Kit and the Genopure Plasmid Maxi Kit.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Ion exchange chromatography is employed by the Genopure Plasmid Midi Kit and the Genopure Plasmid Maxi Kit.

Ion exchange chromatography is employed by the Genopure Plasmid Midi Kit and the Genopure Plasmid Maxi Kit.

Ion exchange chromatography is employed by the Genopure Plasmid Midi Kit and the Genopure Plasmid Maxi Kit.

Ion exchange chromatography is employed by the Genopure Plasmid Midi Kit and the Genopure Plasmid Maxi Kit.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service