Skip to Content
Merck
All Photos(1)

Key Documents

ZTC18S

Millipore

ZipTip® Pipette Tips

C18 resin, bed volume 0.6 μL, tip volume 10 μL

Synonym(s):

Disposable Pipette Tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
eCl@ss:
32033590
NACRES:
NB.24
bed volume:
0.6 μL
thickness:
15 μm
tip I.D.:
3.375 mm
tip L:
31 mm
tip O.D.:
5.8 mm
tip volume:
10 μL
sterility:
non-sterile
feature:
holdup volume 0.2 μL
manufacturer/tradename:
ZipTip®

material

polypropylene

Quality Level

sterility

non-sterile

feature

holdup volume 0.2 μL

manufacturer/tradename

ZipTip®

parameter

4-70  °C temperature

technique(s)

analytical sample preparation: suitable
fractionation: suitable
protein purification: suitable (concentration)

bed volume

0.6 μL

thickness

15 μm

tip I.D.

3.375 mm

tip L

31 mm

tip O.D.

5.8 mm

tip volume

10 μL

pore size

200 Å pore size

pH

1.5-13.5
2-12(for 24 hour exposure)

capacity

5 μg binding capacity(typically; when used with saturating amounts of analyte)

shipped in

ambient

Related Categories

Application

Sample Prep Pipette Tips are suitable for desalting and concentrating peptides or small proteins.
They are also suitable for mass spectrometry sample preparation.

Features and Benefits

  • Single-step desalting, concentration, and purification
  • Fractionate complex samples for more meaningful data
  • Ideal for peptides, proteins, nucleic acids, and more
  • No dead volume for maximum recovery
  • Eliminates time-consuming chromatography

Other Notes

Automation Compatible: No

Legal Information

ZIPTIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Choo Hock Tan et al.
Comparative biochemistry and physiology. Part D, Genomics & proteomics, 43, 101006-101006 (2022-06-20)
The Red-headed Krait (Bungarus flaviceps) is a medically important venomous snake species in Southeast Asia, while there is no specific antivenom available for its envenoming. This study investigated the venom composition through a decomplexation proteomic approach, and examined the immunoreactivity
Sha Liu et al.
Journal of proteome research, 21(7), 1589-1602 (2022-06-18)
Aberrant protein N-glycosylation is a cancer hallmark, which has great potential for cancer detection. However, large-scale and in-depth analysis of N-glycosylation remains challenging because of its high heterogeneity, complexity, and low abundance. Human saliva is an attractive diagnostic body fluid
Mark A Jackson et al.
Frontiers in plant science, 10, 602-602 (2019-06-04)
The backbone cyclic and disulfide bridged sunflower trypsin inhibitor-1 (SFTI-1) peptide is a proven effective scaffold for a range of peptide therapeutics. For production at laboratory scale, solid phase peptide synthesis techniques are widely used, but these synthetic approaches are
Luis Ariel Espinosa et al.
Analytical and bioanalytical chemistry, 413(30), 7559-7585 (2021-11-06)
Subunit vaccines based on the receptor-binding domain (RBD) of the spike protein of SARS-CoV-2 provide one of the most promising strategies to fight the COVID-19 pandemic. The detailed characterization of the protein primary structure by mass spectrometry (MS) is mandatory
Chenchang Bao et al.
Journal of proteomics, 226, 103891-103891 (2020-07-07)
Lysine crotonylation (Kcr) is a recently-discovered type of post-translational modification. Although Kcr has been reported in many species, little is known about this process in crustaceans. In this study, pan anti-lysine crotonylation antibody enrichment and high-resolution liquid chromatogram-mass spectrometry analysis

Articles

Improve your mass spectra with strategies to optimize sample preparation in protein and small molecule mass spectrometry.

Improve your mass spectra with strategies to optimize sample preparation in protein and small molecule mass spectrometry.

Improve your mass spectra with strategies to optimize sample preparation in protein and small molecule mass spectrometry.

Improve your mass spectra with strategies to optimize sample preparation in protein and small molecule mass spectrometry.

Related Content

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service