Skip to Content
Merck
All Photos(1)

Key Documents

MXQTV0KT3

Millipore

Milliflex® Quantum Universal Kit

Includes Milliflex® GSWG funnels, parameter 100 mL, pore size 0.22 μm, (White gridded membrane)

Synonym(s):

Fluorescence-based rapid detection system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
SB.56

material

mixed cellulose esters (MCE)

parameter

100 mL

volume

47 mm

pore size

0.22 μm

General description

Milliflex® Quantum Universal Kit is a fluorescence-based technology for the quantitative and rapid detection of microorganisms in filterable samples. The universal enzymatic fluorescent staining is non-destructive, allowing the identification of the contaminant following a positive result. Therefore, the Milliflex® Quantum system is a fast and reliable alternative for the detection of microorganisms. The kit includes fluorescent reagents and Milliflex® GSWG 100 mL funnels (0.22 μm membrane, white gridded).

Application

The Milliflex® Quantum Universal Kit is used for rapid bioburden testing in filtrable samples, including in-process and final product samples (pharmaceuticals and water). The product is also used in industrial QC and water monitoring.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service