MMP000489
Milliflex® Oasis Funnel
Individually Packed, for use with Milliflex Oasis® systems, gridded white mixed cellulose esters (MCE) filter membrane, pore size 0.45 μm
About This Item
Recommended Products
material
gridded white mixed cellulose esters (MCE) filter membrane
polypropylene body
styrene-acrylonitrile (SAN) body
Quality Level
sterility
sterile; ethylene oxide treated
product line
Milliflex Oasis®
feature
ready-to-use
parameter
100 mL max. working volume
pore size
0.45 μm
application(s)
bioburden testing
cosmetics
pharmaceutical
water monitoring
compatibility
for use with Milliflex Oasis® systems
Looking for similar products? Visit Product Comparison Guide
General description
- 100ml funnel.
- White gridded MCE filter membrane.
The sealed integral membrane filter eliminates the need for membrane manipulation and handling, thus minimizing a major source of contamination and thereby improves bioburden testing reliability. The sealed membrane prevents the bypass phenomenon often found with other filtration systems.
The Milliflex® filtration possesses almost twice the surface area of standard membranes enabling faster filtration and the funnel can be easily detached from the membrane without the use of any tools. The convex shape of the membrane after filtration demonstrates the integrity of the membrane and prevents air bubbles from being trapped when placed on culture media.
Application
Features and Benefits
- Effortless liquid level control, due to the fully transparent material
- Limited liquid residues after filtration, due to the ultra-hydrophobic property of the material
- Effortless funnel and membrane separation
- Reduced risk of secondary contamination, due to the new hinged lid which allows the device to be closed during filtration
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.
Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.
Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.
Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service