Skip to Content
Merck
All Photos(2)

Key Documents

MMMP2MAG-79K

Millipore

MILLIPLEX® Mouse MMP Magnetic Bead Panel 2 - Immunology Multiplex Assay

TIMP Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple TIMP biomarkers in mouse serum, plasma and cell culture samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

standard curve range: 12-50,000 pg/mL
(proMMP-9)

standard curve range: 5-20,000 pg/mL
(MMP-12)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The MMPs (matrix metalloproteinases), a family of zinc proteases responsible for the breakdown of extracellular matrix (ECM), play a key role in normal physiological processes, such as embryonic development and tissue morphogenesis, tissue and bone remodeling, wound healing, and angiogenesis. These processes rely on MMPs′ role in the cleavage of cell surface receptors, the release of apoptotic ligands, cell proliferation and differentiation, and chemokine activity modulation. Similar in structure, MMPs are synthesized and secreted as inactive pro-enzymes that require proteolytic cleavage for activation. This process can be mediated by serine proteases or other MMPs. An increase in MMP expression occurs in response to a wide range of stimuli, including adhesion molecules, growth factors, cytokines, and hormones. Regulation of MMP activity is controlled primarily by TIMPs (tissue inhibitors of metalloproteinases). Therefore, disruption of the MMP/TIMP balance can result in autoimmune disease, arthritis, cardiovascular disease, and tumor growth and metastasis.

MILLIPLEX® Mouse MMP Bead Panel 2, MMMP2MAG-79K, is a 2 plex kit to be used for the simultaneous quantification of the following analytes in serum/plasma samples: proMMP-9 and MMP-12.

Panel Type: Cytokines/Chemokines

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: proMMP-9, MMP-12
  • Recommended Sample type: serum or plasma
  • Recommended Sample dilution: 1:2
  • Assay Run Time: overnight or one day
  • Research Category: Inflammation & Immunology

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: See protocol for sensitive’s of individual analytes

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Noël et al.
Frontiers in physiology, 12, 704401-704401 (2021-12-17)
Matrix metalloproteinase-12 (Mmp12) is upregulated by cigarette smoke (CS) and plays a critical role in extracellular matrix remodeling, a key mechanism involved in physiological repair processes, and in the pathogenesis of emphysema, asthma, and lung cancer. While cigarette smoking is

Related Content

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Immunology multiplex assays like MILLIPLEX® allow simultaneous investigation of immune biomarkers, saving time and resources in research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service