Skip to Content
Merck
All Photos(1)

Key Documents

MAGE1MAG-25K

Millipore

MILLIPLEX® Mouse Aging Magnetic Bead Panel 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

species reactivity

mouse

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Mice are widely used models for both aging and senescence because of their similarity to humans. Aging associated physiological changes and the pathological consequences of these changes make the elderly population vulnerable to many chronic diseases, such as cancer, heart disease, arthritis, osteoporosis, and cognitive decline. Many of these conditions have been modelled in mice. Over the years, intensive research of model organisms has provided fundamental evidence that aging can be delayed, and longevity extended. These findings gave rise to a new era in aging research aiming to elucidate the underlying mechanisms that drive the aging process.The MILLIPLEX® MAP Mouse Aging Magnetic Bead Panel 1 covers multifaceted biomarkers ofaging-related conditions. Monitoring these novel biomarkers helps researchers to furtherunderstand the complex biological process which may lead to new therapies postponing age-related diseases.The MILLIPLEX® MAP Mouse Aging Magnetic Bead Panel 1 is an 8-plex kit to be used for the simultaneous quantification of any or all of the following analytes in mouse serum, plasma and cell/tissue culture samples: C-C Motif Chemokine 27 (CTACK), Fibroblast Growth Factor 21 (FGF21), Progonadoliberin (GnRH), Interleukin-6 (IL-6), Protein Jagged-1 (Jagged1), Leptin, Neurogenic Locus Notch Homolog Protein 1 (Notch1), and Tumor Necrosis Factor Alpha (TNFα).

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.

Application

For optimal sensitivity, use an overnight (16-18 hour) incubation. Serum and plasma samples require (recommend) a 1:2 dilution with Assay Buffer. Tissue culture supernatants may be run neat or appropriately diluted in control medium. Each well requires 25 μL of sample, standard or control.Analytes available:CTACK/CCL27, FGF-21, GnRH, IL-6, Jag1, Leptin, Notch1, TNFα.

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Multiplex immunoassays like MILLIPLEX® Human Myokine Panel enable comprehensive myokine analysis, providing insights into related diseases.

Multiplex immunoassays like MILLIPLEX® Human Myokine Panel enable comprehensive myokine analysis, providing insights into related diseases.

Multiplex immunoassays like MILLIPLEX® Human Myokine Panel enable comprehensive myokine analysis, providing insights into related diseases.

Multiplex immunoassays like MILLIPLEX® Human Myokine Panel enable comprehensive myokine analysis, providing insights into related diseases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service