Skip to Content
Merck
All Photos(3)

Key Documents

MABS1907

Sigma-Aldrich

Anti-Ketohexokinase Isoform C Antibody, clone 5F1-3F5

clone 5F1-3F5, from mouse

Synonym(s):

EC: 2.7.1.3, Hepatic fructokinase, Fructokinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F1-3F5, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... KHK(3795)

General description

Ketohexokinase (UniProt: P50053; also known as EC: 2.7.1.3, Hepatic fructokinase) is encoded by the KHK gene (Gene ID: 3795) in human. Ketohexokinase is a homodimeric cytosolic enzyme in that catalyzes the conversion of fructose to fructose 1 phosphate. It is most abundant in the liver, where it may constitute up to 0.6% of the total liver protein. It has also been reported in kidney, spleen, gut, and pancreas. Very low levels are also found in adrenal, muscle, brain, and eyes. It has multiple fructose binding sites. There are two major isoforms of this enzyme ketofructokinase C and ketofructokinase A. Due to its high Km (~ 28 mM), fructose is preferentially metabolized by the C isoform. Ketohexokinase -C is unique among sugar kinases in that its metabolism of fructose is associated with a rapid depletion of intracellular ATP. Unlike glucokinase, in which excessive phosphorylation of glucose is prevented by a negative feedback system, the metabolism of fructose by ketohexokinase-C results in rapid phosphorylation with a fall in intracellular phosphate and ATP. If the ATP levels fall significantly then the protein synthesis is transiently stopped and cells may develop features that are consistent with ischemia. Mutations in KHK gene has been linked to fructosuria, a clinically benign condition that is characterized nu the incomplete metabolism of fructose in the liver, leading to its urinary excretion. (Ref.: Diggle, CP et al., (2009). J. Histochem. Cytochem. 57(8): 763-774; Hayward, BE., and Bonthron, DT (1998). Eur. J. Biochem. 257(1); 85-91).

Specificity

Clone 5F1-3F5 specifically detects isoform C of human ketohexokinase.

Immunogen

A full-length human recombinant Ketohexokinase, isoform C.

Application

Anti-Ketohexokinase Isoform C, clone 5F1-3F5, Cat. No. MABS1907, is a mouse monoclonal antibody that detectsKetohexokinase and has been tested for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: 2 µg/mL from a represenative lot detected Ketohexokinase Isoform C in HEK293 cells expressing HA-tagged KHK-C.

Western Blotting Analysis: 1 µg/mL from a represenative lot detected Ketohexokinase Isoform C in human liver tissue lysate.

Quality

Evaluated by Western Blotting in human kidney tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Ketohexokinase Isoform C in human kidney tissue lysate.

Target description

~33 kDa observed; 32.52 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service