Skip to Content
Merck
All Photos(2)

Key Documents

MABC97

Sigma-Aldrich

Anti-S1PR4/EDG6 Antibody, clone 16B9.1

clone 16B9.1, from mouse

Synonym(s):

Sphingosine 1-phosphate receptor 4, S1P receptor 4, S1P4, Endothelial differentiation G-protein coupled receptor 6, Sphingosine 1-phosphate receptor Edg-6, S1P receptor Edg-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

16B9.1, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... S1PR4(8698)

General description

Sphingosine 1-phosphate receptor 4 (S1PR4), otherwise known as EDG6, is present, in varying expression levels, in the adult lymph node, bone marrow, and thymus, as well as fetal liver, lung, and thymus. It is activated by the lysosphingolipid sphingosine 1-phosphate, and is coupled to Gi and G12/13 G proteins and via their various effector molecules, SIP4 regulates cell proliferation, differentiation, motility, and migration.

Immunogen

GST-tagged recombinant protein corresponding to human S1PR4/EDG6.

Application

Detect S1PR4/EDG6 using this Anti-S1PR4/EDG6 Antibody, clone 16B9.1 validated for use in Western Blotting & IHC.
Immunohistochemistry Analysis: A 1:300 dilution from a representative lot detected S1PR4/EDG6 in human red bone marrow and human tonsil tissues.

Quality

Evaluated by Western Blot Analysis in rat spleen tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected S1PR4/EDG6 in 10 µg of rat spleen tissue lysate.

Target description

~42 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qun Zhu et al.
The FEBS journal, 281(12), 2861-2870 (2014-05-07)
It has been reported that the effect of inflammatory cytokines on β-cell destruction in type 1 diabetes is concentration-dependent. However, the underlying mechanisms remain unclear. In the present study, we found that a high concentration of cytokines promoted apoptosis in
Xiao-Jing Sun et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 32(8), 1313-1322 (2017-02-17)
C5a plays a crucial role in anti-neutrophil cytoplasmic antibody (ANCA)-mediated neutrophil recruitment and activation. Our previous studies found that the interaction between sphingosine-1-phosphate (S1P) and C5a plays an important role in the ANCA-mediated activation of neutrophils. In the current study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service